Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460432_g1
          See other EIF1 GE Assays ›
          Gene Symbol:
          Eif1
          Gene Name:
          eukaryotic translation initiation factor 1
          Gene Aliases:
          Sui1-rs1
          Chromosome Location:
          Chr.10: 88227455 - 88229546 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460432_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eif1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105837.1 NP_001099307.1 2 - 3 346 168
          GenBank mRNA BC159406.1 - 2 - 3 332 168
          FQ212688.1 - 2 - 3 338 168
          FQ213689.1 - 2 - 3 195 168
          FQ217644.1 - 1 - 2 104 168
          FQ228394.1 - 2 - 3 340 168

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001626.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          translational initiation
          dosage compensation by inactivation of X chromosome
          RNA binding
          translation initiation factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P053252 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022644 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61877 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047730 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047730 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047730 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P397441 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10875373 - -
          Affymetrix Rat Gene 1.0 ST Array 10738172 - -
          Affymetrix Rat Gene 1.1 ST Array 10875372 - -
          Affymetrix Rat Gene 1.1 ST Array 10738170 - -
          Rat Genome 230 2.0 Array 1371378_PM_at - -
          Rat Genome 230 2.0 Array 1388310_PM_at - -
          Rat Genome 230 2.0 Array 1388310_at - -
          Rat Genome 230 2.0 Array 1371378_at - -
          Affymetrix HT Rat Focus Array 1388310_at - -
          Affymetrix HT Rat Focus Array 1371378_PM_at - -
          Affymetrix HT Rat Focus Array 1371378_at - -
          Affymetrix HT Rat Focus Array 1388310_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000014378.rn.1 TC1000001626.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000014380.rn.1 TC1000001626.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000014383.rn.1 TC1000001626.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000014376.rn.1 TC1000001626.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000014381.rn.1 TC1000001626.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000014379.rn.1 TC1000001626.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000014382.rn.1 TC1000001626.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000014370.rn.1 TC1000001626.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000014374.rn.1 TC1000001626.rn.1 RTA TC
          Rat Expression Set 230 1388310_at - -
          Rat Expression Set 230 1371378_at - -
          Rat Expression Set 230 1371378_PM_at - -
          Rat Expression Set 230 1388310_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10738170 - -
          Rat Genome U34 Set rc_AI009501_at - -
          Rat Genome U34 Set rc_AI232021_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21199689 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370359 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362317 - -
          RatRef-12 Expression BeadChip ILMN_54714 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc