Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01461174_m1
          See other MGAT4B GE Assays ›
          Gene Symbol:
          Mgat4b
          Gene Name:
          mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B
          Gene Aliases:
          RGD1562836
          Chromosome Location:
          Chr.10: 35726501 - 35736338 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01461174_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mgat4b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127533.1 NP_001121005.1 1 - 2 359 77
          XM_006246250.2 XP_006246312.1 1 - 2 402 77
          GenBank mRNA BC166514.1 - 1 - 2 341 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000520.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein N-linked glycosylation
          acetylglucosaminyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P19656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23482 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19656 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19656 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P19656 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19656 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17674 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733236 - -
          Affymetrix Rat Gene 1.0 ST Array 10733232 - -
          Affymetrix Rat Gene 1.0 ST Array 10733240 - -
          Affymetrix Rat Gene 1.0 ST Array 10733235 - -
          Affymetrix Rat Gene 1.0 ST Array 10733231 - -
          Affymetrix Rat Gene 1.0 ST Array 10733227 - -
          Affymetrix Rat Gene 1.0 ST Array 10733230 - -
          Affymetrix Rat Gene 1.0 ST Array 10733228 - -
          Affymetrix Rat Gene 1.0 ST Array 10733238 - -
          Affymetrix Rat Gene 1.0 ST Array 10733234 - -
          Affymetrix Rat Gene 1.0 ST Array 10733233 - -
          Affymetrix Rat Gene 1.0 ST Array 10733229 - -
          Affymetrix Rat Gene 1.0 ST Array 10733226 - -
          Affymetrix Rat Gene 1.0 ST Array 10733237 - -
          Affymetrix Rat Gene 1.0 ST Array 10733239 - -
          Affymetrix Rat Gene 1.1 ST Array 10733225 - -
          Rat Genome 230 2.0 Array 1389279_at - -
          Rat Genome 230 2.0 Array 1373560_at - -
          Rat Genome 230 2.0 Array 1373560_PM_at - -
          Rat Genome 230 2.0 Array 1389279_PM_at - -
          Affymetrix HT Rat Focus Array 1373560_PM_at - -
          Affymetrix HT Rat Focus Array 1373560_at - -
          Affymetrix Rat Clariom S Assay PSR1000004299.rn.1 TC1000000520.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004302.rn.1 TC1000000520.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000004300.rn.1 TC1000000520.rn.1 RTA TC
          Rat Expression Set 230 1373560_at - -
          Rat Expression Set 230 1373560_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733225 - -
          Rat Genome U34 Set rc_AI233172_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22075432 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365502 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0