Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01463018_g1
          See other EMG1 GE Assays ›
          Gene Symbol:
          Emg1
          Gene Name:
          EMG1 N1-specific pseudouridine methyltransferase
          Gene Aliases:
          Grcc2f
          Chromosome Location:
          Chr.4: 157222366 - 157230647 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01463018_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Emg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107888.2 NP_001101358.2 5 - 6 752 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003729.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          blastocyst development
          nucleologenesis
          rRNA methylation
          rRNA base methylation
          RNA binding
          rRNA binding
          rRNA (pseudouridine) methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P748981 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P115306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37771 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P748981 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115306 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115306 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P748981 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P115306 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P748981 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P748981 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87998 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10865453 - -
          Affymetrix Rat Gene 1.0 ST Array 10865456 - -
          Affymetrix Rat Gene 1.0 ST Array 10865455 - -
          Affymetrix Rat Gene 1.0 ST Array 10865458 - -
          Affymetrix Rat Gene 1.0 ST Array 10865454 - -
          Affymetrix Rat Gene 1.0 ST Array 10865457 - -
          Affymetrix Rat Gene 1.1 ST Array 10865452 - -
          Rat Genome 230 2.0 Array 1377797_PM_at - -
          Rat Genome 230 2.0 Array 1377797_at - -
          Affymetrix HT Rat Focus Array 1377797_at - -
          Affymetrix HT Rat Focus Array 1377797_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400026350.rn.1 TC0400003729.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026351.rn.1 TC0400003729.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026352.rn.1 TC0400003729.rn.1 RTA TC
          Rat Expression Set 230 1377797_at - -
          Rat Expression Set 230 1377797_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10865452 - -
          Rat Genome U34 Set rc_AI071470_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21936340 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374749 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0