Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01463253_m1
          See other CD9 GE Assays ›
          Gene Symbol:
          Cd9
          Gene Name:
          CD9 molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 157977163 - 158010091 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01463253_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053018.1 NP_444177.1 1 - 2 130 98
          GenBank mRNA BC161804.1 - 1 - 2 105 98
          CF111078.1 - 1 - 2 139 98
          FQ212497.1 - 1 - 2 154 98
          FQ213039.1 - 1 - 2 179 98
          FQ213302.1 - 1 - 2 148 98
          FQ215636.1 - 1 - 2 142 98
          FQ216402.1 - 1 - 2 179 98
          FQ216770.1 - 1 - 2 166 98
          FQ219799.1 - 1 - 2 153 98
          FQ220476.1 - 1 - 2 154 98
          FQ224453.1 - 1 - 2 154 98
          FQ229965.1 - 1 - 2 201 98
          FQ230141.1 - 1 - 2 216 98
          X76489.1 - 1 - 2 69 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003767.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          fusion of sperm to egg plasma membrane involved in single fertilization
          brain development
          negative regulation of cell population proliferation
          response to water deprivation
          oligodendrocyte development
          paranodal junction assembly
          integrin binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P462771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463508 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17723 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90224 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P462771 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P462771 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P462771 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P462771 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96493 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10865722 - -
          Affymetrix Rat Gene 1.0 ST Array 10865720 - -
          Affymetrix Rat Gene 1.0 ST Array 10865718 - -
          Affymetrix Rat Gene 1.0 ST Array 10865723 - -
          Affymetrix Rat Gene 1.0 ST Array 10865716 - -
          Affymetrix Rat Gene 1.0 ST Array 10865717 - -
          Affymetrix Rat Gene 1.0 ST Array 10865721 - -
          Affymetrix Rat Gene 1.0 ST Array 10865719 - -
          Affymetrix Rat Gene 1.1 ST Array 10865715 - -
          Rat Genome 230 2.0 Array 1371499_PM_at - -
          Rat Genome 230 2.0 Array 1371499_at - -
          Affymetrix HT Rat Focus Array 1371499_at - -
          Affymetrix HT Rat Focus Array 1371499_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400026833.rn.1 TC0400003767.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026834.rn.1 TC0400003767.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026835.rn.1 TC0400003767.rn.1 RTA TC
          Rat Expression Set 230 1371499_PM_at - -
          Rat Expression Set 230 1371499_at - -
          Affymetrix Rat Gene 1.0 ST Array 10865715 - -
          Rat Genome U34 Set X76489cds_at - -
          Rat Genome U34 Set X76489cds_g_at - -
          Rat Genome U34 Set rc_AI012389_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22072229 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0