Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01463732_m1
          See other FAM134C GE Assays ›
          Gene Symbol:
          Fam134c
          Gene Name:
          family with sequence similarity 134, member C
          Gene Aliases:
          RGD1561189
          Chromosome Location:
          Chr.10: 89006595 - 89030595 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01463732_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam134c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135804.1 NP_001129276.1 5 - 6 616 77
          GenBank mRNA BC166581.1 - 5 - 6 616 77
          FQ212358.1 - 5 - 6 733 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003819.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915818 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147623 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52663 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062433 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062433 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062433 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P558204 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56638 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81616 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22313 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10747569 - -
          Affymetrix Rat Gene 1.0 ST Array 10747567 - -
          Affymetrix Rat Gene 1.0 ST Array 10747568 - -
          Affymetrix Rat Gene 1.0 ST Array 10747570 - -
          Affymetrix Rat Gene 1.0 ST Array 10747566 - -
          Affymetrix Rat Gene 1.0 ST Array 10747565 - -
          Affymetrix Rat Gene 1.0 ST Array 10747572 - -
          Affymetrix Rat Gene 1.0 ST Array 10747574 - -
          Affymetrix Rat Gene 1.0 ST Array 10747573 - -
          Affymetrix Rat Gene 1.0 ST Array 10747571 - -
          Affymetrix Rat Gene 1.1 ST Array 10747564 - -
          Rat Genome 230 2.0 Array 1376087_PM_at - -
          Rat Genome 230 2.0 Array 1396131_PM_at - -
          Rat Genome 230 2.0 Array 1396131_at - -
          Rat Genome 230 2.0 Array 1376087_at - -
          Affymetrix HT Rat Focus Array 1376087_at - -
          Affymetrix HT Rat Focus Array 1396131_at - -
          Affymetrix HT Rat Focus Array 1376087_PM_at - -
          Affymetrix HT Rat Focus Array 1396131_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000034321.rn.1 TC1000003819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034325.rn.1 TC1000003819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034319.rn.1 TC1000003819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034324.rn.1 TC1000003819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034322.rn.1 TC1000003819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034320.rn.1 TC1000003819.rn.1 RTA TC
          Rat Expression Set 230 1376087_PM_at - -
          Rat Expression Set 230 1376087_at - -
          Rat Expression Set 230 1396131_PM_at - -
          Rat Expression Set 230 1396131_at - -
          Affymetrix Rat Gene 1.0 ST Array 10747564 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21657324 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349217 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0