Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01465808_m1
          See other OLFM4 GE Assays ›
          Gene Symbol:
          Olfm4
          Gene Name:
          olfactomedin 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 62384262 - 62429450 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01465808_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfm4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106052.1 NP_001099522.1 4 - 5 752 110
          XM_008770863.1 XP_008769085.1 4 - 5 1579 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000914.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of necrotic cell death
          regulation of apoptotic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of phagocytosis
          negative regulation of immune response
          protein homooligomerization
          positive regulation of substrate adhesion-dependent cell spreading
          regulation of neutrophil extravasation
          protein homodimerization activity
          cadherin binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P487468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424183 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38784 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91861 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75247 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10330 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66720 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P487468 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P487468 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P487468 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P487468 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69438 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781724 - -
          Affymetrix Rat Gene 1.0 ST Array 10781723 - -
          Affymetrix Rat Gene 1.0 ST Array 10781726 - -
          Affymetrix Rat Gene 1.0 ST Array 10781729 - -
          Affymetrix Rat Gene 1.0 ST Array 10781725 - -
          Affymetrix Rat Gene 1.1 ST Array 10781722 - -
          Affymetrix Rat Clariom S Assay PSR1500006336.rn.1 TC1500000914.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10781722 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21592827 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc