Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01466080_m1
          See other LOXL2 GE Assays ›
          Gene Symbol:
          Loxl2
          Gene Name:
          lysyl oxidase-like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 51276022 - 51365238 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01466080_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Loxl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106047.2 NP_001099517.2 10 - 11 2117 76
          GenBank mRNA BC168900.1 - 10 - 11 2117 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000759.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          epithelial to mesenchymal transition
          endothelial cell proliferation
          sprouting angiogenesis
          transcription, DNA-templated
          cellular protein modification process
          receptor-mediated endocytosis
          histone modification
          protein deamination
          collagen fibril organization
          positive regulation of chondrocyte differentiation
          endothelial cell migration
          negative regulation of transcription, DNA-templated
          response to copper ion
          chromatin binding
          transcription corepressor activity
          protein-lysine 6-oxidase activity
          scavenger receptor activity
          copper ion binding
          methylated histone binding
          oligosaccharide binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P105225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64616 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28776 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156864 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161173 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109684 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64748 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129858 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P105225 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P105225 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P105225 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P105225 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72411 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23909 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781315 - -
          Affymetrix Rat Gene 1.0 ST Array 10781317 - -
          Affymetrix Rat Gene 1.0 ST Array 10781311 - -
          Affymetrix Rat Gene 1.0 ST Array 10781310 - -
          Affymetrix Rat Gene 1.0 ST Array 10781305 - -
          Affymetrix Rat Gene 1.0 ST Array 10781313 - -
          Affymetrix Rat Gene 1.0 ST Array 10781309 - -
          Affymetrix Rat Gene 1.0 ST Array 10781312 - -
          Affymetrix Rat Gene 1.0 ST Array 10781306 - -
          Affymetrix Rat Gene 1.0 ST Array 10781316 - -
          Affymetrix Rat Gene 1.0 ST Array 10781314 - -
          Affymetrix Rat Gene 1.0 ST Array 10781318 - -
          Affymetrix Rat Gene 1.0 ST Array 10781308 - -
          Affymetrix Rat Gene 1.0 ST Array 10781319 - -
          Affymetrix Rat Gene 1.1 ST Array 10781304 - -
          Rat Genome 230 2.0 Array 1391450_PM_at - -
          Rat Genome 230 2.0 Array 1380474_PM_at - -
          Rat Genome 230 2.0 Array 1380474_at - -
          Rat Genome 230 2.0 Array 1391450_at - -
          Affymetrix HT Rat Focus Array 1380474_at - -
          Affymetrix HT Rat Focus Array 1391450_PM_at - -
          Affymetrix HT Rat Focus Array 1380474_PM_at - -
          Affymetrix HT Rat Focus Array 1391450_at - -
          Affymetrix Rat Clariom S Assay PSR1500005258.rn.1 TC1500000759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005259.rn.1 TC1500000759.rn.1 RTA TC
          Rat Expression Set 230 1391450_at - -
          Rat Expression Set 230 1380474_PM_at - -
          Rat Expression Set 230 1391450_PM_at - -
          Rat Expression Set 230 1380474_at - -
          Affymetrix Rat Gene 1.0 ST Array 10781304 - -
          Rat Genome U34 Set rc_AI180353_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351352 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline