Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01466149_m1
          See other SERP2 GE Assays ›
          Gene Symbol:
          Serp2
          Gene Name:
          stress-associated endoplasmic reticulum protein family member 2
          Gene Aliases:
          RGD1566076
          Chromosome Location:
          Chr.15: 58687784 - 58712015 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01466149_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109104.1 NP_001102574.1 3 - 4 541 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500002099.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P144016 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P684315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381101 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80082 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111317 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144016 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P684315 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144016 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P684315 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P144016 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P684315 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P684315 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103980 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10785357 - -
          Affymetrix Rat Gene 1.0 ST Array 10785358 - -
          Affymetrix Rat Gene 1.0 ST Array 10785356 - -
          Affymetrix Rat Gene 1.1 ST Array 10785355 - -
          Rat Genome 230 2.0 Array 1374772_PM_at - -
          Rat Genome 230 2.0 Array 1374772_at - -
          Affymetrix HT Rat Focus Array 1374772_at - -
          Affymetrix HT Rat Focus Array 1374772_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500014454.rn.1 TC1500002099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014460.rn.1 TC1500002099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014456.rn.1 TC1500002099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014457.rn.1 TC1500002099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014459.rn.1 TC1500002099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014458.rn.1 TC1500002099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014455.rn.1 TC1500002099.rn.1 RTA TC
          Rat Expression Set 230 1374772_PM_at - -
          Rat Expression Set 230 1374772_at - -
          Affymetrix Rat Gene 1.0 ST Array 10785355 - -
          Rat Genome U34 Set rc_AI045755_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358457 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0