Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01466227_g1
          See other CLN5 GE Assays ›
          Gene Symbol:
          Cln5 RGD1564125
          Gene Name:
          ceroid-lipofuscinosis, neuronal 5
          similar to Cln5 protein
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.15: 93634815 - 93644146 on Build Rnor_6.0
          Chromosome Location:
          Chr.2: 66600817 - 66615026 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01466227_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cln5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191689.1 NP_001178618.1 283 75
          Gene Symbol: RGD1564125
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008760838.1 XP_008759060.1 2 451 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001061.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          signal peptide processing
          lysosome organization
          lysosomal lumen acidification
          brain development
          visual perception
          biological_process
          neurogenesis
          glycosylation
          molecular_function
          mannose binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P443101 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P468723 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P116430 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116430 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P443101 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P468723 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P443101 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P468723 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116430 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P116430 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P443101 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P468723 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P443101 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781883 - -
          Rat Genome 230 2.0 Array 1375971_at - -
          Rat Genome 230 2.0 Array 1375971_PM_at - -
          Affymetrix HT Rat Focus Array 1375971_PM_at - -
          Affymetrix HT Rat Focus Array 1375971_at - -
          Affymetrix Rat Clariom S Assay PSR1500006986.rn.1 TC1500001061.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500006988.rn.1 TC1500001061.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500006992.rn.1 TC1500001061.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500006987.rn.1 TC1500001061.rn.1 RTA TC
          Rat Expression Set 230 1375971_at - -
          Rat Expression Set 230 1375971_PM_at - -
          Rat Genome U34 Set rc_AI011208_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21978152 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650480 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372804 - -
          RatRef-12 Expression BeadChip ILMN_52518 - -
          RatRef-12 Expression BeadChip ILMN_50925 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0