Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01468822_m1
          See other SOS2 GE Assays ›
          Gene Symbol:
          Sos2
          Gene Name:
          son of sevenless homolog 2 (Drosophila)
          Gene Aliases:
          Sos1
          Chromosome Location:
          Chr.6: 91885292 - 92008059 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01468822_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sos2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135561.1 NP_001129033.1 18 - 19 2961 68
          XM_006240182.2 XP_006240244.1 18 - 19 3100 68
          XM_006240184.2 XP_006240246.1 15 - 16 3506 68
          XM_008764721.1 XP_008762943.1 17 - 18 3004 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002603.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          B cell homeostasis
          lymphocyte homeostasis
          small GTPase mediated signal transduction
          regulation of T cell differentiation in thymus
          regulation of T cell proliferation
          positive regulation of GTPase activity
          positive regulation of small GTPase mediated signal transduction
          regulation of pro-B cell differentiation
          molecular_function
          DNA binding
          guanyl-nucleotide exchange factor activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P579569 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P142430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822993 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103363 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12023 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152954 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57248 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78044 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106652 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66332 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P579569 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142430 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142430 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P579569 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P579569 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P142430 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P579569 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P559085 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15943 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102483 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10890330 - -
          Affymetrix Rat Gene 1.0 ST Array 10890336 - -
          Affymetrix Rat Gene 1.0 ST Array 10890332 - -
          Affymetrix Rat Gene 1.0 ST Array 10890326 - -
          Affymetrix Rat Gene 1.0 ST Array 10890316 - -
          Affymetrix Rat Gene 1.0 ST Array 10890321 - -
          Affymetrix Rat Gene 1.0 ST Array 10890334 - -
          Affymetrix Rat Gene 1.0 ST Array 10890329 - -
          Affymetrix Rat Gene 1.0 ST Array 10890325 - -
          Affymetrix Rat Gene 1.0 ST Array 10890323 - -
          Affymetrix Rat Gene 1.0 ST Array 10890322 - -
          Affymetrix Rat Gene 1.0 ST Array 10890324 - -
          Affymetrix Rat Gene 1.0 ST Array 10890327 - -
          Affymetrix Rat Gene 1.0 ST Array 10890328 - -
          Affymetrix Rat Gene 1.0 ST Array 10890320 - -
          Affymetrix Rat Gene 1.0 ST Array 10890333 - -
          Affymetrix Rat Gene 1.0 ST Array 10890335 - -
          Affymetrix Rat Gene 1.0 ST Array 10890315 - -
          Affymetrix Rat Gene 1.0 ST Array 10890337 - -
          Affymetrix Rat Gene 1.0 ST Array 10890318 - -
          Affymetrix Rat Gene 1.0 ST Array 10890319 - -
          Affymetrix Rat Gene 1.0 ST Array 10890331 - -
          Affymetrix Rat Gene 1.0 ST Array 10890317 - -
          Affymetrix Rat Gene 1.1 ST Array 10890314 - -
          Affymetrix Rat Gene 1.1 ST Array 10890301 - -
          Rat Genome 230 2.0 Array 1376489_at - -
          Rat Genome 230 2.0 Array 1376489_PM_at - -
          Affymetrix HT Rat Focus Array 1376489_PM_at - -
          Affymetrix HT Rat Focus Array 1376489_at - -
          Affymetrix Rat Clariom S Assay PSR0600018402.rn.1 TC0600002603.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600018403.rn.1 TC0600002603.rn.1 RTA TC
          Rat Expression Set 230 1376489_at - -
          Rat Expression Set 230 1376489_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10890314 - -
          Rat Genome U34 Set rc_AI044857_at - -
          Rat Genome U34 Set rc_AI228913_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20990981 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358348 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0