Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01470685_m1
          See other LSM11 GE Assays ›
          Gene Symbol:
          Lsm11
          Gene Name:
          LSM11, U7 small nuclear RNA associated
          Gene Aliases:
          RGD1565523
          Chromosome Location:
          Chr.10: 31023561 - 31041741 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01470685_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lsm11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001173384.1 NP_001166855.1 2 - 3 605 64
          GenBank mRNA BC158836.1 - 2 - 3 605 64
          FQ223124.1 - 2 - 3 641 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002638.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA 3'-end processing by stem-loop binding and cleavage
          positive regulation of G1/S transition of mitotic cell cycle
          U7 snRNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P049497 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049496 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049496 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P049496 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66238 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100563 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742233 - -
          Affymetrix Rat Gene 1.0 ST Array 10742231 - -
          Affymetrix Rat Gene 1.0 ST Array 10742234 - -
          Affymetrix Rat Gene 1.0 ST Array 10742232 - -
          Affymetrix Rat Gene 1.1 ST Array 10742230 - -
          Rat Genome 230 2.0 Array 1382557_PM_at - -
          Rat Genome 230 2.0 Array 1382557_at - -
          Rat Genome 230 2.0 Array 1397874_at - -
          Rat Genome 230 2.0 Array 1397874_PM_at - -
          Affymetrix HT Rat Focus Array 1397874_PM_at - -
          Affymetrix HT Rat Focus Array 1397874_at - -
          Affymetrix Rat Clariom S Assay PSR1000023458.rn.1 TC1000002638.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000023459.rn.1 TC1000002638.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000023460.rn.1 TC1000002638.rn.1 RTA TC
          Rat Expression Set 230 1397874_PM_at - -
          Rat Expression Set 230 1397874_at - -
          Rat Expression Set 230 1382557_PM_at - -
          Rat Expression Set 230 1382557_at - -
          Affymetrix Rat Gene 1.0 ST Array 10742230 - -
          Rat Genome U34 Set rc_AA946491_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357459 - -
          RatRef-12 Expression BeadChip ILMN_64825 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0