Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01470738_g1
          See other ACTG1 GE Assays ›
          Gene Symbol:
          Actg1 LOC100361457
          Gene Name:
          actin, gamma 1
          actin, gamma 1 propeptide-like
          Gene Aliases:
          Actg
          -
          Chromosome Location:
          Chr.10: 109518433 - 109521288 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 75643057 - 75644964 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01470738_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Actg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127449.1 NP_001120921.1 1 - 2 65 77
          GenBank mRNA CK595239.1 - 80 77
          FQ216158.1 - 1 - 2 61 77
          Gene Symbol: LOC100361457
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002729196.3 XP_002729242.2 1 62 77

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001127449.1, XM_002729196.3 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          sarcomere organization
          response to calcium ion
          platelet aggregation
          molecular_function
          structural constituent of cytoskeleton
          ATP binding
          ubiquitin protein ligase binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P090244 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P378624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184460 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P090244 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P378624 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P090244 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P378624 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P090244 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P378624 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P700674 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770889 - -
          Affymetrix Rat Gene 1.0 ST Array 10705873 - -
          Affymetrix Rat Gene 1.0 ST Array 10749618 - -
          Affymetrix Rat Gene 1.0 ST Array 10749614 - -
          Affymetrix Rat Gene 1.0 ST Array 10803654 - -
          Affymetrix Rat Gene 1.0 ST Array 10765091 - -
          Affymetrix Rat Gene 1.0 ST Array 10749617 - -
          Affymetrix Rat Gene 1.0 ST Array 10749613 - -
          Affymetrix Rat Gene 1.0 ST Array 10749616 - -
          Affymetrix Rat Gene 1.0 ST Array 10749615 - -
          Affymetrix Rat Gene 1.0 ST Array 10837538 - -
          Affymetrix Rat Gene 1.1 ST Array 10705872 - -
          Affymetrix Rat Gene 1.1 ST Array 10770888 - -
          Affymetrix Rat Gene 1.1 ST Array 10803653 - -
          Affymetrix Rat Gene 1.1 ST Array 10749612 - -
          Affymetrix Rat Gene 1.1 ST Array 10765090 - -
          Affymetrix Rat Gene 1.1 ST Array 10837537 - -
          Rat Genome 230 2.0 Array 1371327_PM_a_at - -
          Rat Genome 230 2.0 Array 1371327_a_at - -
          Rat Genome 230 2.0 Array 1384581_PM_at - -
          Rat Genome 230 2.0 Array 1384581_at - -
          Affymetrix HT Rat Focus Array 1384581_PM_at - -
          Affymetrix HT Rat Focus Array 1371327_a_at - -
          Affymetrix HT Rat Focus Array 1371327_PM_a_at - -
          Affymetrix HT Rat Focus Array 1384581_at - -
          Rat Expression Set 230 1371327_PM_a_at - -
          Rat Expression Set 230 1371327_a_at - -
          Rat Expression Set 230 1384581_PM_at - -
          Rat Expression Set 230 1384581_at - -
          Affymetrix Rat Gene 1.0 ST Array 10837537 - -
          Affymetrix Rat Gene 1.0 ST Array 10705872 - -
          Affymetrix Rat Gene 1.0 ST Array 10770888 - -
          Affymetrix Rat Gene 1.0 ST Array 10749612 - -
          Affymetrix Rat Gene 1.0 ST Array 10803653 - -
          Affymetrix Rat Gene 1.0 ST Array 10765090 - -
          Rat Genome U34 Set X52815cds_f_at - -
          Rat Genome U34 Set rc_AA964496_s_at - -
          Rat Genome U34 Set rc_AI104357_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22334636 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364152 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362269 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-998b77544-nf2nv:80/100.66.134.75:80.
          git-commit: efd2b0e08ab99238c7874a09efb06372d741ac0a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc