Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01473516_m1
          See other RGD1562284 GE Assays ›
          Gene Symbol:
          RGD1562284
          Gene Name:
          similar to Glutaminyl-peptide cyclotransferase precursor (QC)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 1657114 - 1689575 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01473516_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1562284
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134557.2 NP_001128029.1 5 - 6 802 147
          XM_006239632.2 XP_006239694.1 6 - 7 1167 147
          GenBank mRNA FQ228724.1 - 6 - 7 958 147
          FQ229106.1 - 6 - 7 961 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000019.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-pyroglutamic acid biosynthetic process, using glutaminyl-peptide cyclotransferase
          zinc ion binding
          glutaminyl-peptide cyclotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P365569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26097 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125563 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163707 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150333 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P365569 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P365569 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P365569 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P365569 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10882462 - -
          Affymetrix Rat Gene 1.0 ST Array 10882469 - -
          Affymetrix Rat Gene 1.0 ST Array 10882470 - -
          Affymetrix Rat Gene 1.0 ST Array 10882466 - -
          Affymetrix Rat Gene 1.0 ST Array 10882468 - -
          Affymetrix Rat Gene 1.0 ST Array 10882465 - -
          Affymetrix Rat Gene 1.0 ST Array 10882467 - -
          Affymetrix Rat Gene 1.0 ST Array 10882463 - -
          Affymetrix Rat Gene 1.1 ST Array 10882461 - -
          Rat Genome 230 2.0 Array 1377867_at - -
          Rat Genome 230 2.0 Array 1377867_PM_at - -
          Affymetrix HT Rat Focus Array 1377867_at - -
          Affymetrix HT Rat Focus Array 1377867_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600000148.rn.1 TC0600000019.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600000147.rn.1 TC0600000019.rn.1 RTA TC
          Rat Expression Set 230 1377867_at - -
          Rat Expression Set 230 1377867_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10882461 - -
          Rat Genome U34 Set rc_AA859661_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21423688 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650699 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0