Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01473655_g1
          See other NFKBIA GE Assays ›
          Gene Symbol:
          Nfkbia
          Gene Name:
          nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha
          Gene Aliases:
          RL/IF-1
          Chromosome Location:
          Chr.6: 76267227 - 76270457 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01473655_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nfkbia
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105720.2 NP_001099190.2 4 - 5 751 115
          GenBank mRNA BC166886.1 - 4 - 5 703 115
          FQ214065.1 - 2 - 3 199 115
          FQ215246.1 - 4 - 5 703 115
          FQ225350.1 - 4 - 5 698 115
          FQ225877.1 - 4 - 5 701 115
          FQ226288.1 - 4 - 5 701 115
          FQ227283.1 - 4 - 5 701 115
          FQ233200.1 - 4 - 5 701 115
          FQ233557.1 - 4 - 5 703 115
          FQ233627.1 - 4 - 5 689 115
          X63594.1 - 4 - 5 681 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002478.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoplasmic sequestering of NF-kappaB
          regulation of gene expression
          negative regulation of macrophage derived foam cell differentiation
          positive regulation of cholesterol efflux
          negative regulation of lipid storage
          lipopolysaccharide-mediated signaling pathway
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of cellular protein metabolic process
          response to muramyl dipeptide
          response to lipopolysaccharide
          toll-like receptor 4 signaling pathway
          response to muscle stretch
          regulation of cell population proliferation
          cytoplasmic sequestering of transcription factor
          response to exogenous dsRNA
          negative regulation of myeloid cell differentiation
          negative regulation of Notch signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          nucleotide-binding oligomerization domain containing 1 signaling pathway
          nucleotide-binding oligomerization domain containing 2 signaling pathway
          nuclear localization sequence binding
          enzyme binding
          heat shock protein binding
          ubiquitin protein ligase binding
          identical protein binding
          protein-containing complex binding
          NF-kappaB binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0600017680.rn.1 TC0600002478.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600017679.rn.1 TC0600002478.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6bbccd986b-mx52t:80/100.66.130.62:80.
          git-commit: 717389b07c2b9f75290ff02e811d41bb86d4254a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0