Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01474624_m1
          See other LOC100910732 GE Assays ›
          Gene Symbol:
          LOC100910732 Pak2
          Gene Name:
          serine/threonine-protein kinase PAK 2-like
          p21 protein (Cdc42/Rac)-activated kinase 2
          Gene Aliases:
          -
          gamma-PAK
          Chromosome Location:
          Chr.11: 71959063 - 72021248 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 72829407 - 72865815 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01474624_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100910732
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003751066.2 XP_003751114.1 14 - 15 1765 150
          XM_006248473.1 XP_006248535.1 15 - 16 1894 150
          XM_008768778.1 XP_008767000.1 14 - 15 3326 150
          Gene Symbol: Pak2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053306.3 NP_445758.2 1617 150
          GenBank mRNA U19967.1 - 1617 150

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000763.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100910732
          Click here to find siRNAs for Gene Pak2

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          protein phosphorylation
          phosphorylation
          peptidyl-serine phosphorylation
          intracellular signal transduction
          regulation of apoptotic process
          negative regulation of apoptotic process
          protein autophosphorylation
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of protein tyrosine kinase activity
          positive regulation of extrinsic apoptotic signaling pathway
          negative regulation of cysteine-type endopeptidase activity involved in execution phase of apoptosis
          protein serine/threonine kinase activity
          ATP binding
          protein kinase binding
          protein tyrosine kinase activator activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P689755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288575 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77103 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148609 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163424 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17670 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39402 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P689755 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P689755 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P689755 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P689755 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56439 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68541 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24678 - -
          Agilent Rat cDNA Microarray (G4105A) 702436586 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10751675 - -
          Affymetrix Rat Gene 1.0 ST Array 10751682 - -
          Affymetrix Rat Gene 1.0 ST Array 10751676 - -
          Affymetrix Rat Gene 1.0 ST Array 10751685 - -
          Affymetrix Rat Gene 1.0 ST Array 10751678 - -
          Affymetrix Rat Gene 1.0 ST Array 10751677 - -
          Affymetrix Rat Gene 1.0 ST Array 10751673 - -
          Affymetrix Rat Gene 1.0 ST Array 10751681 - -
          Affymetrix Rat Gene 1.0 ST Array 10751674 - -
          Affymetrix Rat Gene 1.0 ST Array 10751686 - -
          Affymetrix Rat Gene 1.0 ST Array 10751679 - -
          Affymetrix Rat Gene 1.0 ST Array 10751680 - -
          Affymetrix Rat Gene 1.0 ST Array 10751684 - -
          Affymetrix Rat Gene 1.0 ST Array 10751683 - -
          Affymetrix Rat Gene 1.1 ST Array 10751671 - -
          Rat Genome 230 2.0 Array 1396267_at - -
          Rat Genome 230 2.0 Array 1393184_PM_at - -
          Rat Genome 230 2.0 Array 1369650_at - -
          Rat Genome 230 2.0 Array 1396267_PM_at - -
          Rat Genome 230 2.0 Array 1388345_PM_at - -
          Rat Genome 230 2.0 Array 1393184_at - -
          Rat Genome 230 2.0 Array 1369650_PM_at - -
          Rat Genome 230 2.0 Array 1388345_at - -
          Affymetrix HT Rat Focus Array 1396267_PM_at - -
          Affymetrix HT Rat Focus Array 1369650_PM_at - -
          Affymetrix HT Rat Focus Array 1369650_at - -
          Affymetrix HT Rat Focus Array 1396267_at - -
          Affymetrix Rat Clariom S Assay PSR1100004208.rn.1 TC1100000763.rn.1 RTA TC
          Rat Expression Set 230 1369650_PM_at - -
          Rat Expression Set 230 1369650_at - -
          Rat Expression Set 230 1396267_at - -
          Rat Expression Set 230 1396267_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10751671 - -
          Rat Genome U34 Set U35345_s_at - -
          Rat Genome U34 Set U35345_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21975446 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372206 - -
          RatRef-12 Expression BeadChip ILMN_58549 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0