Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100652.1 | NP_001094122.1 | 8 - 9 | 894 | 71 | |
GenBank mRNA | BC091408.1 | - | 7 - 8 | 666 | 71 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |