Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01475302_g1
          See other LOC103694586 GE Assays ›
          Gene Symbol:
          LOC103694586 Aifm1
          Gene Name:
          apoptosis-inducing factor 1, mitochondrial-like
          apoptosis-inducing factor, mitochondrion-associated 1
          Gene Aliases:
          -
          Aif, Pdcd8
          Chromosome Location:
          Chr.X|NW_007906396: 11858 - 20329 on Build Rnor_6.0
          Chromosome Location:
          Chr.X: 135304063 - 135343062 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01475302_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103694586
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008773721.1 XP_008771943.1 2 - 3 235 93
          Gene Symbol: Aifm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031356.1 NP_112646.1 15 - 16 1848 93
          GenBank mRNA AB041723.1 - 15 - 16 1864 93
          AF262320.1 - 15 - 16 1770 93
          AF375656.1 - 15 - 16 1774 93
          BC072697.1 - 15 - 16 1806 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002450.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC103694586
          Click here to find siRNAs for Gene Aifm1

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of cell death
          neuron differentiation
          mitochondrial respiratory chain complex I assembly
          cell redox homeostasis
          neuron apoptotic process
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          regulation of apoptotic DNA fragmentation
          DNA binding
          NAD(P)H oxidase H2O2-forming activity
          oxidoreductase activity, acting on NAD(P)H
          protein dimerization activity
          FAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199649 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44024 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74511 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062801 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062801 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062801 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14387 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700531818 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51894 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18946 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89979 - -
          Agilent Rat cDNA Microarray (G4105A) 700531818 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10939597 - -
          Affymetrix Rat Gene 1.0 ST Array 10939603 - -
          Affymetrix Rat Gene 1.0 ST Array 10939612 - -
          Affymetrix Rat Gene 1.0 ST Array 10939598 - -
          Affymetrix Rat Gene 1.0 ST Array 10939601 - -
          Affymetrix Rat Gene 1.0 ST Array 10939614 - -
          Affymetrix Rat Gene 1.0 ST Array 10939599 - -
          Affymetrix Rat Gene 1.0 ST Array 10939610 - -
          Affymetrix Rat Gene 1.0 ST Array 10939596 - -
          Affymetrix Rat Gene 1.0 ST Array 10939608 - -
          Affymetrix Rat Gene 1.0 ST Array 10939613 - -
          Affymetrix Rat Gene 1.0 ST Array 10939609 - -
          Affymetrix Rat Gene 1.0 ST Array 10939602 - -
          Affymetrix Rat Gene 1.0 ST Array 10939600 - -
          Affymetrix Rat Gene 1.0 ST Array 10939605 - -
          Affymetrix Rat Gene 1.0 ST Array 10939606 - -
          Affymetrix Rat Gene 1.0 ST Array 10939607 - -
          Affymetrix Rat Gene 1.0 ST Array 10939604 - -
          Affymetrix Rat Gene 1.1 ST Array 10939595 - -
          Rat Genome 230 2.0 Array 1370321_at - -
          Rat Genome 230 2.0 Array 1370321_PM_at - -
          Affymetrix HT Rat Focus Array 1370321_PM_at - -
          Affymetrix HT Rat Focus Array 1370321_at - -
          Affymetrix Rat Clariom S Assay PSR0X00015302.rn.1 TC0X00002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00015304.rn.1 TC0X00002450.rn.1 RTA TC
          Rat Expression Set 230 1370321_PM_at - -
          Rat Expression Set 230 1370321_at - -
          Affymetrix Rat Gene 1.0 ST Array 10939595 - -
          Rat Genome U34 Set rc_AA891591_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22393003 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368612 - -
          RatRef-12 Expression BeadChip ILMN_64240 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0