Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01476533_g1
          See other CATSPERG1 GE Assays ›
          Gene Symbol:
          Catsperg1
          Gene Name:
          cation channel, sperm-associated, gamma 1
          Gene Aliases:
          Catsperg, RGD1310942
          Chromosome Location:
          Chr.1: 88131287 - 88162616 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01476533_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Catsperg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170340.1 NP_001163811.1 27 - 28 3132 101
          XM_006228722.2 XP_006228784.1 27 - 28 3718 101
          XM_008759124.1 XP_008757346.1 27 - 28 3751 101
          XM_008759125.1 XP_008757347.1 27 - 28 3733 101
          XM_008759126.1 XP_008757348.1 26 - 27 3643 101
          XM_008759127.1 XP_008757349.1 26 - 27 2949 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005122.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P054408 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P054403 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P372640 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054403 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P372640 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054403 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054403 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P372640 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P372640 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10720448 - -
          Affymetrix Rat Gene 1.0 ST Array 10720456 - -
          Affymetrix Rat Gene 1.0 ST Array 10720451 - -
          Affymetrix Rat Gene 1.0 ST Array 10720445 - -
          Affymetrix Rat Gene 1.0 ST Array 10720455 - -
          Affymetrix Rat Gene 1.0 ST Array 10720450 - -
          Affymetrix Rat Gene 1.0 ST Array 10720457 - -
          Affymetrix Rat Gene 1.0 ST Array 10720454 - -
          Affymetrix Rat Gene 1.0 ST Array 10720446 - -
          Affymetrix Rat Gene 1.0 ST Array 10720453 - -
          Affymetrix Rat Gene 1.0 ST Array 10720458 - -
          Affymetrix Rat Gene 1.0 ST Array 10720447 - -
          Affymetrix Rat Gene 1.0 ST Array 10720449 - -
          Affymetrix Rat Gene 1.0 ST Array 10720452 - -
          Affymetrix Rat Gene 1.1 ST Array 10720437 - -
          Affymetrix Rat Clariom S Assay PSR0100040976.rn.1 TC0100005122.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040977.rn.1 TC0100005122.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040978.rn.1 TC0100005122.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040979.rn.1 TC0100005122.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040975.rn.1 TC0100005122.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10720437 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20889881 - -
          ABI Rat Genome Survey Microarray 21932808 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357983 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360045 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0