Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01479552_m1
          See other TRPM5 GE Assays ›
          Gene Symbol:
          Trpm5
          Gene Name:
          transient receptor potential cation channel, subfamily M, member 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 216256661 - 216281500 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01479552_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpm5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191896.1 NP_001178825.1 4 - 5 657 81
          XM_008760160.1 XP_008758382.1 6 - 7 2864 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006996.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transmembrane transport
          sodium ion transmembrane transport
          sensory perception of taste
          potassium ion transmembrane transport
          calcium activated cation channel activity
          potassium channel activity
          sodium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P041541 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041541 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041541 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P041541 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P375125 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0100055313.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055317.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055321.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055342.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055314.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055326.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055334.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055336.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055337.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055340.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055343.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055328.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055329.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055333.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055323.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055332.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055344.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055315.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055320.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055324.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055327.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055335.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055318.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055331.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055339.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055319.rn.1 TC0100006996.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055330.rn.1 TC0100006996.rn.1 RTA TC
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21009907 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369731 - -
          RatRef-12 Expression BeadChip ILMN_67044 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0