Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01482039_m1
          See other CARKD GE Assays ›
          Gene Symbol:
          Carkd
          Gene Name:
          carbohydrate kinase domain containing
          Gene Aliases:
          RGD1562691
          Chromosome Location:
          Chr.16: 83326228 - 83342688 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01482039_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Carkd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108402.1 NP_001101872.1 9 - 10 863 146
          XM_006253465.2 XP_006253527.1 9 - 10 990 146
          GenBank mRNA FQ225763.1 - 825 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600002047.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          nicotinamide nucleotide metabolic process
          molecular_function
          ATP binding
          ATP-dependent NAD(P)H-hydrate dehydratase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P189837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413820 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15459 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132467 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P189837 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P189837 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P189837 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P189837 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39304 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792952 - -
          Affymetrix Rat Gene 1.0 ST Array 10792951 - -
          Affymetrix Rat Gene 1.0 ST Array 10792949 - -
          Affymetrix Rat Gene 1.0 ST Array 10792953 - -
          Affymetrix Rat Gene 1.0 ST Array 10792947 - -
          Affymetrix Rat Gene 1.0 ST Array 10792948 - -
          Affymetrix Rat Gene 1.0 ST Array 10792950 - -
          Affymetrix Rat Gene 1.0 ST Array 10792946 - -
          Affymetrix Rat Gene 1.1 ST Array 10792943 - -
          Rat Genome 230 2.0 Array 1379759_PM_at - -
          Rat Genome 230 2.0 Array 1391583_at - -
          Rat Genome 230 2.0 Array 1379759_at - -
          Rat Genome 230 2.0 Array 1391583_PM_at - -
          Affymetrix HT Rat Focus Array 1391583_at - -
          Affymetrix HT Rat Focus Array 1379759_at - -
          Affymetrix HT Rat Focus Array 1391583_PM_at - -
          Affymetrix HT Rat Focus Array 1379759_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600015282.rn.1 TC1600002047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015284.rn.1 TC1600002047.rn.1 RTA TC
          Rat Expression Set 230 1379759_PM_at - -
          Rat Expression Set 230 1379759_at - -
          Rat Expression Set 230 1391583_PM_at - -
          Rat Expression Set 230 1391583_at - -
          Affymetrix Rat Gene 1.0 ST Array 10792943 - -
          Rat Genome U34 Set rc_AA944188_at - -
          Rat Genome U34 Set H32870_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21519992 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364980 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0