Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483035_m1
          See other MCM6 GE Assays ›
          Gene Symbol:
          Mcm6
          Gene Name:
          minichromosome maintenance complex component 6
          Gene Aliases:
          Mcmd6
          Chromosome Location:
          Chr.13: 45042882 - 45068073 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483035_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mcm6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017287.1 NP_058983.1 7 - 8 1190 69
          GenBank mRNA BC166822.1 - 7 - 8 1110 69
          CD373304.1 - 1 - 2 102 69
          U17565.1 - 1 - 2 140 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001348.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          DNA unwinding involved in DNA replication
          DNA replication initiation
          DNA binding
          DNA helicase activity
          single-stranded DNA binding
          helicase activity
          ATP binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P276338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48497 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105880 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P276338 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P276338 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P276338 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P276338 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42214 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99476 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767347 - -
          Affymetrix Rat Gene 1.0 ST Array 10767342 - -
          Affymetrix Rat Gene 1.0 ST Array 10767345 - -
          Affymetrix Rat Gene 1.0 ST Array 10767355 - -
          Affymetrix Rat Gene 1.0 ST Array 10767344 - -
          Affymetrix Rat Gene 1.0 ST Array 10767348 - -
          Affymetrix Rat Gene 1.0 ST Array 10767339 - -
          Affymetrix Rat Gene 1.0 ST Array 10767354 - -
          Affymetrix Rat Gene 1.0 ST Array 10767346 - -
          Affymetrix Rat Gene 1.0 ST Array 10767350 - -
          Affymetrix Rat Gene 1.0 ST Array 10767352 - -
          Affymetrix Rat Gene 1.0 ST Array 10767349 - -
          Affymetrix Rat Gene 1.0 ST Array 10767351 - -
          Affymetrix Rat Gene 1.0 ST Array 10767341 - -
          Affymetrix Rat Gene 1.0 ST Array 10767343 - -
          Affymetrix Rat Gene 1.0 ST Array 10767340 - -
          Affymetrix Rat Gene 1.0 ST Array 10767353 - -
          Affymetrix Rat Gene 1.1 ST Array 10767338 - -
          Rat Genome 230 2.0 Array 1371074_a_at - -
          Rat Genome 230 2.0 Array 1371074_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371074_a_at - -
          Affymetrix HT Rat Focus Array 1371074_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1300008833.rn.1 TC1300001348.rn.1 RTA TC
          Rat Expression Set 230 1371074_a_at - -
          Rat Expression Set 230 1371074_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10767338 - -
          Rat Genome U34 Set rc_AI639082_s_at - -
          Rat Genome U34 Set U17565_at - -
          Rat Genome U34 Set U17565_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22054034 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356031 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0