Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483384_g1
          See other CSRP1 GE Assays ›
          Gene Symbol:
          Csrp1
          Gene Name:
          cysteine and glycine-rich protein 1
          Gene Aliases:
          Csrp
          Chromosome Location:
          Chr.13: 52553843 - 52575054 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483384_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csrp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017148.2 NP_058844.1 5 - 6 544 84
          GenBank mRNA BC062407.1 - 5 - 6 544 84
          U09567.1 - 5 - 6 539 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000339.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csrp1

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          platelet aggregation
          RNA binding
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32859 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69689 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11838 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11838 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11838 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11838 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32116 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74763 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10764224 - -
          Affymetrix Rat Gene 1.0 ST Array 10764225 - -
          Affymetrix Rat Gene 1.0 ST Array 10764226 - -
          Affymetrix Rat Gene 1.0 ST Array 10764227 - -
          Affymetrix Rat Gene 1.0 ST Array 10764222 - -
          Affymetrix Rat Gene 1.0 ST Array 10764223 - -
          Affymetrix Rat Gene 1.1 ST Array 10764221 - -
          Rat Genome 230 2.0 Array 1370057_at - -
          Rat Genome 230 2.0 Array 1370057_PM_at - -
          Affymetrix HT Rat Focus Array 1370057_PM_at - -
          Affymetrix HT Rat Focus Array 1370057_at - -
          Affymetrix Rat Clariom S Assay PSR1300002233.rn.1 TC1300000339.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300002234.rn.1 TC1300000339.rn.1 RTA TC
          Rat Expression Set 230 1370057_at - -
          Rat Expression Set 230 1370057_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10764221 - -
          Rat Genome U34 Set rc_AI234146_at - -
          Rat Genome U34 Set rc_AI234100_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22349130 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370050 - -
          RatRef-12 Expression BeadChip ILMN_54969 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline