Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483752_m1
          See other TIPRL GE Assays ›
          Gene Symbol:
          Tiprl
          Gene Name:
          TOR signaling pathway regulator
          Gene Aliases:
          RGD1310442
          Chromosome Location:
          Chr.13: 83478966 - 83504448 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483752_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tiprl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109667.1 NP_001103137.1 2 - 3 330 124
          GenBank mRNA BC128780.1 - 2 - 3 330 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001823.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          DNA damage checkpoint signaling
          negative regulation of phosphoprotein phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P041253 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P041254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69002 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78943 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51441 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53462 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041253 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041254 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041254 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041253 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P041253 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P041254 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P488840 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6290 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769512 - -
          Affymetrix Rat Gene 1.0 ST Array 10769515 - -
          Affymetrix Rat Gene 1.0 ST Array 10769514 - -
          Affymetrix Rat Gene 1.0 ST Array 10769516 - -
          Affymetrix Rat Gene 1.0 ST Array 10769511 - -
          Affymetrix Rat Gene 1.0 ST Array 10769513 - -
          Affymetrix Rat Gene 1.0 ST Array 10769510 - -
          Affymetrix Rat Gene 1.1 ST Array 10769509 - -
          Rat Genome 230 2.0 Array 1372397_PM_at - -
          Rat Genome 230 2.0 Array 1372397_at - -
          Affymetrix HT Rat Focus Array 1372397_PM_at - -
          Affymetrix HT Rat Focus Array 1372397_at - -
          Affymetrix Rat Clariom S Assay PSR1300012886.rn.1 TC1300001823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012891.rn.1 TC1300001823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012887.rn.1 TC1300001823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012890.rn.1 TC1300001823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012892.rn.1 TC1300001823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012893.rn.1 TC1300001823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012885.rn.1 TC1300001823.rn.1 RTA TC
          Rat Expression Set 230 1372397_at - -
          Rat Expression Set 230 1372397_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769509 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21086368 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650013 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0