Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483826_m1
          See other CENPL GE Assays ›
          Gene Symbol:
          Cenpl
          Gene Name:
          centromere protein L
          Gene Aliases:
          RGD1311868
          Chromosome Location:
          Chr.13: 78886111 - 78901025 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483826_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cenpl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033061.2 NP_001028233.2 5 - 6 1460 83
          NM_001244761.1 NP_001231690.1 4 - 5 1374 83
          GenBank mRNA BC079467.1 - 5 - 6 1460 83
          BC089893.1 - 4 - 5 1374 83
          BC103642.1 - 5 - 6 1453 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000606.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cenpl

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P480361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45810 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101379 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123248 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115454 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157856 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81322 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15651 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480361 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P480361 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P480361 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98908 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74025 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10765045 - -
          Affymetrix Rat Gene 1.0 ST Array 10765049 - -
          Affymetrix Rat Gene 1.0 ST Array 10765048 - -
          Affymetrix Rat Gene 1.0 ST Array 10765046 - -
          Affymetrix Rat Gene 1.0 ST Array 10765050 - -
          Affymetrix Rat Gene 1.0 ST Array 10765047 - -
          Affymetrix Rat Gene 1.1 ST Array 10765044 - -
          Rat Genome 230 2.0 Array 1398608_PM_at - -
          Rat Genome 230 2.0 Array 1398608_at - -
          Affymetrix HT Rat Focus Array 1398608_at - -
          Affymetrix HT Rat Focus Array 1398608_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300004194.rn.1 TC1300000606.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004196.rn.1 TC1300000606.rn.1 RTA TC
          Rat Expression Set 230 1398608_PM_at - -
          Rat Expression Set 230 1398608_at - -
          Affymetrix Rat Gene 1.0 ST Array 10765044 - -
          Rat Genome U34 Set rc_AI104608_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22184092 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357608 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0