Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483880_m1
          See other MFSD4 GE Assays ›
          Gene Symbol:
          Mfsd4
          Gene Name:
          major facilitator superfamily domain containing 4
          Gene Aliases:
          RGD1566090
          Chromosome Location:
          Chr.13: 48807720 - 48848997 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483880_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mfsd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109069.1 NP_001102539.1 9 - 10 1648 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001392.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          transmembrane transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P015366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015366 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015366 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P015366 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30470 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767568 - -
          Affymetrix Rat Gene 1.0 ST Array 10767573 - -
          Affymetrix Rat Gene 1.0 ST Array 10767566 - -
          Affymetrix Rat Gene 1.0 ST Array 10767572 - -
          Affymetrix Rat Gene 1.0 ST Array 10767571 - -
          Affymetrix Rat Gene 1.0 ST Array 10767575 - -
          Affymetrix Rat Gene 1.0 ST Array 10767574 - -
          Affymetrix Rat Gene 1.0 ST Array 10767569 - -
          Affymetrix Rat Gene 1.0 ST Array 10767576 - -
          Affymetrix Rat Gene 1.0 ST Array 10767570 - -
          Affymetrix Rat Gene 1.1 ST Array 10767565 - -
          Affymetrix Rat Clariom S Assay PSR1300009221.rn.1 TC1300001392.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10767565 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22167599 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353003 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0