Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01484156_m1
          See other SPC25 GE Assays ›
          Gene Symbol:
          Spc25
          Gene Name:
          SPC25, NDC80 kinetochore complex component
          Gene Aliases:
          RGD1307811, Spbc25
          Chromosome Location:
          Chr.3: 55438534 - 55451934 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01484156_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spc25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009654.1 NP_001009654.1 6 - 7 630 87
          XM_006234288.2 XP_006234350.1 6 - 7 636 87
          GenBank mRNA BC088213.1 - 6 - 7 630 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003250.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spc25

          Gene Ontology Categories:

          Function(s)

          mitotic cell cycle
          mitotic spindle organization
          chromosome segregation
          cell division

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P050303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109432 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P050303 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P050303 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P050303 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P253108 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9272 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10845970 - -
          Affymetrix Rat Gene 1.0 ST Array 10845975 - -
          Affymetrix Rat Gene 1.0 ST Array 10845973 - -
          Affymetrix Rat Gene 1.0 ST Array 10845972 - -
          Affymetrix Rat Gene 1.0 ST Array 10845976 - -
          Affymetrix Rat Gene 1.0 ST Array 10845974 - -
          Affymetrix Rat Gene 1.0 ST Array 10845971 - -
          Affymetrix Rat Gene 1.1 ST Array 10845969 - -
          Rat Genome 230 2.0 Array 1389668_PM_at - -
          Rat Genome 230 2.0 Array 1389668_at - -
          Affymetrix HT Rat Focus Array 1389668_at - -
          Affymetrix HT Rat Focus Array 1389668_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300025249.rn.1 TC0300003250.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300025247.rn.1 TC0300003250.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300025248.rn.1 TC0300003250.rn.1 RTA TC
          Rat Expression Set 230 1389668_PM_at - -
          Rat Expression Set 230 1389668_at - -
          Affymetrix Rat Gene 1.0 ST Array 10845969 - -
          Rat Genome U34 Set rc_AI045594_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21384927 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365457 - -
          RatRef-12 Expression BeadChip ILMN_53432 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0