Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01484645_m1
          See other TMC2 GE Assays ›
          Gene Symbol:
          Tmc2
          Gene Name:
          transmembrane channel-like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 122720769 - 122790100 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01484645_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106510.1 NP_001099980.1 18 - 19 2573 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001710.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of ion transmembrane transport
          detection of mechanical stimulus involved in sensory perception of sound
          vestibular reflex
          calcium ion transmembrane transport
          voltage-gated calcium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115907 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29768 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177598 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106971 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111078 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017675 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017675 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017675 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P147381 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45000 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75190 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66827 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10839840 - -
          Affymetrix Rat Gene 1.0 ST Array 10839843 - -
          Affymetrix Rat Gene 1.0 ST Array 10839854 - -
          Affymetrix Rat Gene 1.0 ST Array 10839853 - -
          Affymetrix Rat Gene 1.0 ST Array 10839850 - -
          Affymetrix Rat Gene 1.0 ST Array 10839842 - -
          Affymetrix Rat Gene 1.0 ST Array 10839848 - -
          Affymetrix Rat Gene 1.0 ST Array 10839841 - -
          Affymetrix Rat Gene 1.0 ST Array 10839851 - -
          Affymetrix Rat Gene 1.0 ST Array 10839849 - -
          Affymetrix Rat Gene 1.0 ST Array 10839845 - -
          Affymetrix Rat Gene 1.0 ST Array 10839855 - -
          Affymetrix Rat Gene 1.0 ST Array 10839847 - -
          Affymetrix Rat Gene 1.0 ST Array 10839852 - -
          Affymetrix Rat Gene 1.0 ST Array 10839844 - -
          Affymetrix Rat Gene 1.0 ST Array 10839856 - -
          Affymetrix Rat Gene 1.0 ST Array 10839846 - -
          Affymetrix Rat Gene 1.1 ST Array 10839838 - -
          Affymetrix Rat Clariom S Assay PSR0300012261.rn.1 TC0300001710.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10839838 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21782674 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359170 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370597 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0