Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01486354_m1
          See other LOC100911313 GE Assays ›
          Gene Symbol:
          LOC100911313 Rmdn3
          Gene Name:
          regulator of microtubule dynamics protein 3-like
          regulator of microtubule dynamics 3
          Gene Aliases:
          -
          Fam82a2, Fam82c, Ptpip51, RGD1308697, Rmd-3
          Chromosome Location:
          Chr.3: 112605090 - 112616301 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 110943843 - 110964449 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01486354_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100911313
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008762136.1 XP_008760358.1 9 - 10 1137 136
          Gene Symbol: Rmdn3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014046.2 NP_001014068.1 12 - 13 1457 136
          GenBank mRNA BC082081.1 - 12 - 13 1419 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003971.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100911313
          Click here to find siRNAs for Gene Rmdn3

          Gene Ontology Categories:

          Function(s)

          cellular calcium ion homeostasis
          apoptotic process
          cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P365882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150604 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29446 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P365882 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P365882 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P365882 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P365882 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81907 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81711 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10848511 - -
          Affymetrix Rat Gene 1.0 ST Array 10848507 - -
          Affymetrix Rat Gene 1.0 ST Array 10848505 - -
          Affymetrix Rat Gene 1.0 ST Array 10848508 - -
          Affymetrix Rat Gene 1.0 ST Array 10848504 - -
          Affymetrix Rat Gene 1.0 ST Array 10848510 - -
          Affymetrix Rat Gene 1.0 ST Array 10848509 - -
          Affymetrix Rat Gene 1.0 ST Array 10848512 - -
          Affymetrix Rat Gene 1.0 ST Array 10848513 - -
          Affymetrix Rat Gene 1.0 ST Array 10848514 - -
          Affymetrix Rat Gene 1.0 ST Array 10848515 - -
          Affymetrix Rat Gene 1.0 ST Array 10848506 - -
          Affymetrix Rat Gene 1.1 ST Array 10848503 - -
          Rat Genome 230 2.0 Array 1372302_PM_at - -
          Rat Genome 230 2.0 Array 1372302_at - -
          Affymetrix HT Rat Focus Array 1372302_at - -
          Affymetrix HT Rat Focus Array 1372302_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300030064.rn.1 TC0300003971.rn.1 RTA TC
          Rat Expression Set 230 1372302_PM_at - -
          Rat Expression Set 230 1372302_at - -
          Affymetrix Rat Gene 1.0 ST Array 10848503 - -
          Rat Genome U34 Set rc_AA800224_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21294428 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367951 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc