Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsper2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012220.1 | NP_001012220.1 | 6 - 7 | 1060 | 95 | |
XM_006234860.2 | XP_006234922.1 | 6 - 7 | 986 | 95 | ||
XM_008762187.1 | XP_008760409.1 | 6 - 7 | 742 | 95 | ||
GenBank mRNA | BC079422.1 | - | 6 - 7 | 1060 | 95 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |