Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01486513_m1
          See other ZFP846 GE Assays ›
          Gene Symbol:
          Zfp846
          Gene Name:
          zinc finger protein 846
          Gene Aliases:
          RGD1564241, Zfp426l2, Znf426, Znf846
          Chromosome Location:
          Chr.8: 21601112 - 21618389 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01486513_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp846
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134585.2 NP_001128057.1 5 - 6 647 134
          XM_006242645.2 XP_006242707.1 5 - 6 1372 134
          XM_006242647.2 XP_006242709.1 6 - 7 974 134
          XM_006242648.2 XP_006242710.1 6 - 7 1548 134
          XM_008765958.1 XP_008764180.1 4 - 5 1792 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000195.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761583 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16584 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011291 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011291 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011291 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10863 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10908250 - -
          Affymetrix Rat Gene 1.0 ST Array 10908252 - -
          Affymetrix Rat Gene 1.0 ST Array 10908249 - -
          Affymetrix Rat Gene 1.0 ST Array 10908251 - -
          Affymetrix Rat Gene 1.1 ST Array 10908248 - -
          Rat Genome 230 2.0 Array 1391564_PM_at - -
          Rat Genome 230 2.0 Array 1393543_at - -
          Rat Genome 230 2.0 Array 1391564_at - -
          Rat Genome 230 2.0 Array 1393543_PM_at - -
          Affymetrix HT Rat Focus Array 1391564_at - -
          Affymetrix HT Rat Focus Array 1393543_at - -
          Affymetrix HT Rat Focus Array 1391564_PM_at - -
          Affymetrix HT Rat Focus Array 1393543_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800000993.rn.1 TC0800000195.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800000994.rn.1 TC0800000195.rn.1 RTA TC
          Rat Expression Set 230 1393543_at - -
          Rat Expression Set 230 1391564_at - -
          Rat Expression Set 230 1391564_PM_at - -
          Rat Expression Set 230 1393543_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10908248 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21419807 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374390 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-867f8cf58d-sfwzd:80/100.66.130.62:80.
          git-commit: 7a7e1ef32845d1da800a461bcf18a29ea3e3afc0
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix