Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01487035_m1
          See other ZFP426 GE Assays ›
          Gene Symbol:
          Zfp426
          Gene Name:
          zinc finger protein 426
          Gene Aliases:
          Znf426
          Chromosome Location:
          Chr.8: 21477328 - 21511233 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01487035_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp426
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001079943.1 NP_001073412.1 6 - 7 528 105
          XM_006242666.1 XP_006242728.1 5 - 6 535 105
          XM_006242667.2 XP_006242729.1 5 - 6 2623 105
          XM_006242668.2 XP_006242730.1 6 - 7 1929 105
          XM_006242670.1 XP_006242732.1 6 - 7 618 105
          XM_006242671.2 XP_006242733.1 4 - 5 404 105
          XM_006242672.2 XP_006242734.1 5 - 6 2134 105
          XM_006242673.2 XP_006242735.1 4 - 5 1984 105
          XM_008765981.1 XP_008764203.1 5 - 6 2134 105
          XM_008765982.1 XP_008764204.1 5 - 6 471 105
          XM_008765983.1 XP_008764205.1 5 - 6 1779 105
          XM_008765984.1 XP_008764206.1 5 - 6 1735 105
          GenBank mRNA BC129125.1 - 6 - 7 528 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002040.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P480830 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P137297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266591 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169518 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30802 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137297 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480830 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480830 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137297 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P137297 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P480830 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P480830 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95862 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10915330 - -
          Affymetrix Rat Gene 1.0 ST Array 10915331 - -
          Affymetrix Rat Gene 1.0 ST Array 10915329 - -
          Affymetrix Rat Gene 1.0 ST Array 10915327 - -
          Affymetrix Rat Gene 1.0 ST Array 10915326 - -
          Affymetrix Rat Gene 1.0 ST Array 10915332 - -
          Affymetrix Rat Gene 1.0 ST Array 10915325 - -
          Affymetrix Rat Gene 1.0 ST Array 10915328 - -
          Affymetrix Rat Gene 1.1 ST Array 10915324 - -
          Rat Genome 230 2.0 Array 1383670_PM_at - -
          Rat Genome 230 2.0 Array 1383670_at - -
          Affymetrix HT Rat Focus Array 1383670_PM_at - -
          Affymetrix HT Rat Focus Array 1383670_at - -
          Affymetrix Rat Clariom S Assay PSR0800016220.rn.1 TC0800002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800016221.rn.1 TC0800002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800016223.rn.1 TC0800002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800016224.rn.1 TC0800002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800016218.rn.1 TC0800002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800016233.rn.1 TC0800002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800016215.rn.1 TC0800002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800016219.rn.1 TC0800002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800016222.rn.1 TC0800002040.rn.1 RTA TC
          Rat Expression Set 230 1383670_at - -
          Rat Expression Set 230 1383670_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10915324 - -
          Rat Genome U34 Set rc_H33614_at - -
          Rat Genome U34 Set rc_AI045647_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0