Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01488958_g1
          See other SAP18 GE Assays ›
          Gene Symbol:
          Sap18 RGD1561590
          Gene Name:
          Sin3-associated polypeptide 18
          similar to SAP18
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.15: 38069320 - 38073625 on Build Rnor_6.0
          Chromosome Location:
          Chr.10: 89635649 - 89636275 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01488958_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sap18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033685.2 NP_001028857.2 3 - 4 365 83
          GenBank mRNA BC104706.1 - 3 - 4 354 83
          CK357541.1 - 3 - 4 322 83
          FQ212380.1 - 3 - 4 337 83
          FQ223271.1 - 3 - 4 332 83
          FQ234134.1 - 3 - 4 373 83
          Gene Symbol: RGD1561590
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002727811.4 XP_002727857.1 1 388 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000574.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sap18
          Click here to find siRNAs for Gene RGD1561590

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          biological_process
          positive regulation of apoptotic process
          negative regulation of mRNA splicing, via spliceosome
          molecular_function
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1037886 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314738 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1037886 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062585 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062585 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1037886 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062585 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1037886 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1037886 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10780804 - -
          Affymetrix Rat Gene 1.0 ST Array 10780805 - -
          Affymetrix Rat Gene 1.0 ST Array 10780806 - -
          Affymetrix Rat Gene 1.0 ST Array 10738481 - -
          Affymetrix Rat Gene 1.0 ST Array 10780807 - -
          Affymetrix Rat Gene 1.1 ST Array 10738480 - -
          Affymetrix Rat Gene 1.1 ST Array 10780803 - -
          Rat Genome 230 2.0 Array 1392465_PM_at - -
          Rat Genome 230 2.0 Array 1392465_at - -
          Affymetrix HT Rat Focus Array 1392465_at - -
          Affymetrix HT Rat Focus Array 1392465_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500004047.rn.1 TC1500000574.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500004048.rn.1 TC1500000574.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500004049.rn.1 TC1500000574.rn.1 RTA TC
          Rat Expression Set 230 1392465_at - -
          Rat Expression Set 230 1392465_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10780803 - -
          Rat Genome U34 Set rc_AI228030_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649787 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365452 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0