Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01490081_m1
          See other GPBP1 GE Assays ›
          Gene Symbol:
          Gpbp1
          Gene Name:
          GC-rich promoter binding protein 1
          Gene Aliases:
          RGD1305492
          Chromosome Location:
          Chr.2: 43002041 - 43070236 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01490081_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106410.1 NP_001099880.1 10 - 11 2215 89
          XM_006231921.2 XP_006231983.1 11 - 12 2581 89
          XM_006231922.2 XP_006231984.1 10 - 11 2521 89
          XM_006231923.1 XP_006231985.1 7 - 8 922 89
          XM_008760725.1 XP_008758947.1 6 - 7 862 89
          GenBank mRNA BC161907.1 - 9 - 10 1434 89
          FQ211661.1 - 11 - 12 2402 89
          FQ213360.1 - 3 - 4 449 89
          JQ013734.1 - 10 - 11 1301 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002879.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          DNA binding
          DNA-binding transcription factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176836 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9962 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77329 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7441 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157244 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142673 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022649 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022649 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P022649 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1046937 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P130226 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P830214 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41769 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10821256 - -
          Affymetrix Rat Gene 1.0 ST Array 10821260 - -
          Affymetrix Rat Gene 1.0 ST Array 10821259 - -
          Affymetrix Rat Gene 1.0 ST Array 10821258 - -
          Affymetrix Rat Gene 1.0 ST Array 10821265 - -
          Affymetrix Rat Gene 1.0 ST Array 10821263 - -
          Affymetrix Rat Gene 1.0 ST Array 10821261 - -
          Affymetrix Rat Gene 1.0 ST Array 10821267 - -
          Affymetrix Rat Gene 1.0 ST Array 10821262 - -
          Affymetrix Rat Gene 1.0 ST Array 10821257 - -
          Affymetrix Rat Gene 1.0 ST Array 10821264 - -
          Affymetrix Rat Gene 1.1 ST Array 10821266 - -
          Affymetrix Rat Gene 1.1 ST Array 10821255 - -
          Rat Genome 230 2.0 Array 1383099_at - -
          Rat Genome 230 2.0 Array 1388563_PM_at - -
          Rat Genome 230 2.0 Array 1396128_at - -
          Rat Genome 230 2.0 Array 1388563_at - -
          Rat Genome 230 2.0 Array 1396128_PM_at - -
          Rat Genome 230 2.0 Array 1383099_PM_at - -
          Affymetrix HT Rat Focus Array 1388563_at - -
          Affymetrix HT Rat Focus Array 1396128_PM_at - -
          Affymetrix HT Rat Focus Array 1383099_at - -
          Affymetrix HT Rat Focus Array 1396128_at - -
          Affymetrix HT Rat Focus Array 1388563_PM_at - -
          Affymetrix HT Rat Focus Array 1383099_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200020249.rn.1 TC0200002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200020255.rn.1 TC0200002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200020250.rn.1 TC0200002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200020252.rn.1 TC0200002879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200020254.rn.1 TC0200002879.rn.1 RTA TC
          Rat Expression Set 230 1388563_at - -
          Rat Expression Set 230 1388563_PM_at - -
          Rat Expression Set 230 1383099_at - -
          Rat Expression Set 230 1396128_PM_at - -
          Rat Expression Set 230 1396128_at - -
          Rat Expression Set 230 1383099_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10821266 - -
          Affymetrix Rat Gene 1.0 ST Array 10821255 - -
          Rat Genome U34 Set rc_AI060057_at - -
          Rat Genome U34 Set rc_AI072266_at - -
          Rat Genome U34 Set rc_AI169287_at - -
          Rat Genome U34 Set rc_AI177885_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20725356 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369054 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349961 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0