Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01490135_m1
          See other ZFP131 GE Assays ›
          Gene Symbol:
          Zfp131
          Gene Name:
          zinc finger protein 131
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 52503185 - 52532008 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01490135_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp131
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100698.1 NP_001094168.1 5 - 6 1201 89
          XM_006231960.2 XP_006232022.1 5 - 6 1196 89
          XM_006231961.2 XP_006232023.1 6 - 7 1425 89
          XM_006231962.2 XP_006232024.1 6 - 7 1251 89
          XM_006231963.2 XP_006232025.1 6 - 7 1274 89
          XM_006231965.2 XP_006232027.1 6 - 7 1439 89
          XM_008760772.1 XP_008758994.1 6 - 7 1526 89
          GenBank mRNA BC098842.1 - 1 - 2 126 89
          BC161974.1 - 5 - 6 1201 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002986.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          DNA-binding transcription factor activity, RNA polymerase II-specific
          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P064376 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P068187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129537 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064376 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068187 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064376 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068187 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P064376 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P068187 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P300637 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P449522 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10821590 - -
          Affymetrix Rat Gene 1.0 ST Array 10821593 - -
          Affymetrix Rat Gene 1.0 ST Array 10821589 - -
          Affymetrix Rat Gene 1.0 ST Array 10821592 - -
          Affymetrix Rat Gene 1.0 ST Array 10821595 - -
          Affymetrix Rat Gene 1.0 ST Array 10821591 - -
          Affymetrix Rat Gene 1.0 ST Array 10821594 - -
          Affymetrix Rat Gene 1.0 ST Array 10821596 - -
          Affymetrix Rat Gene 1.1 ST Array 10821587 - -
          Rat Genome 230 2.0 Array 1395418_PM_at - -
          Rat Genome 230 2.0 Array 1395418_at - -
          Rat Genome 230 2.0 Array 1393780_at - -
          Rat Genome 230 2.0 Array 1393780_PM_at - -
          Affymetrix HT Rat Focus Array 1395418_at - -
          Affymetrix HT Rat Focus Array 1395418_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200020944.rn.1 TC0200002986.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200020945.rn.1 TC0200002986.rn.1 RTA TC
          Rat Expression Set 230 1395418_at - -
          Rat Expression Set 230 1393780_PM_at - -
          Rat Expression Set 230 1393780_at - -
          Rat Expression Set 230 1395418_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10821587 - -
          Rat Genome U34 Set rc_AA924380_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21908743 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353216 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0