Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01492336_m1
          See other TM7SF3 GE Assays ›
          Gene Symbol:
          Tm7sf3
          Gene Name:
          transmembrane 7 superfamily member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 180913703 - 180951564 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01492336_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tm7sf3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011970.1 NP_001011970.1 11 - 12 1532 73
          GenBank mRNA BC090030.1 - 11 - 12 1532 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400004096.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tm7sf3

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1015425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110755 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1015425 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1015425 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1015425 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1015425 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93176 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70462 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79145 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866948 - -
          Affymetrix Rat Gene 1.0 ST Array 10866952 - -
          Affymetrix Rat Gene 1.0 ST Array 10866955 - -
          Affymetrix Rat Gene 1.0 ST Array 10866947 - -
          Affymetrix Rat Gene 1.0 ST Array 10866945 - -
          Affymetrix Rat Gene 1.0 ST Array 10866949 - -
          Affymetrix Rat Gene 1.0 ST Array 10866954 - -
          Affymetrix Rat Gene 1.0 ST Array 10866946 - -
          Affymetrix Rat Gene 1.0 ST Array 10866950 - -
          Affymetrix Rat Gene 1.0 ST Array 10866953 - -
          Affymetrix Rat Gene 1.0 ST Array 10866951 - -
          Affymetrix Rat Gene 1.0 ST Array 10866956 - -
          Affymetrix Rat Gene 1.1 ST Array 10866943 - -
          Rat Genome 230 2.0 Array 1389609_PM_at - -
          Rat Genome 230 2.0 Array 1389609_at - -
          Affymetrix HT Rat Focus Array 1389609_at - -
          Affymetrix HT Rat Focus Array 1389609_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400029113.rn.1 TC0400004096.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029114.rn.1 TC0400004096.rn.1 RTA TC
          Rat Expression Set 230 1389609_at - -
          Rat Expression Set 230 1389609_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10866943 - -
          Rat Genome U34 Set rc_AI102291_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375613 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376531 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline