Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01492385_m1
          See other TSPAN9 GE Assays ›
          Gene Symbol:
          Tspan9
          Gene Name:
          tetraspanin 9
          Gene Aliases:
          RGD1304740
          Chromosome Location:
          Chr.4: 160530726 - 160725048 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01492385_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tspan9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107890.1 NP_001101360.1 6 - 7 1016 117
          XM_006237494.2 XP_006237556.1 10 - 11 1147 117
          XM_006237495.2 XP_006237557.1 9 - 10 963 117
          XM_006237496.2 XP_006237558.1 9 - 10 1026 117
          XM_006237498.2 XP_006237560.1 8 - 9 846 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003807.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P153062 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P655897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142444 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89943 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13339 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37606 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145570 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P655897 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P655897 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P153062 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P655897 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P192336 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104049 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10865803 - -
          Affymetrix Rat Gene 1.0 ST Array 10865806 - -
          Affymetrix Rat Gene 1.0 ST Array 10865804 - -
          Affymetrix Rat Gene 1.0 ST Array 10865801 - -
          Affymetrix Rat Gene 1.0 ST Array 10865805 - -
          Affymetrix Rat Gene 1.0 ST Array 10865808 - -
          Affymetrix Rat Gene 1.0 ST Array 10865810 - -
          Affymetrix Rat Gene 1.0 ST Array 10865807 - -
          Affymetrix Rat Gene 1.0 ST Array 10865802 - -
          Affymetrix Rat Gene 1.1 ST Array 10865800 - -
          Rat Genome 230 2.0 Array 1371602_at - -
          Rat Genome 230 2.0 Array 1377780_PM_at - -
          Rat Genome 230 2.0 Array 1377780_at - -
          Rat Genome 230 2.0 Array 1371602_PM_at - -
          Affymetrix HT Rat Focus Array 1371602_PM_at - -
          Affymetrix HT Rat Focus Array 1377780_at - -
          Affymetrix HT Rat Focus Array 1371602_at - -
          Affymetrix HT Rat Focus Array 1377780_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400027037.rn.1 TC0400003807.rn.1 RTA TC
          Rat Expression Set 230 1371602_PM_at - -
          Rat Expression Set 230 1371602_at - -
          Rat Expression Set 230 1377780_PM_at - -
          Rat Expression Set 230 1377780_at - -
          Affymetrix Rat Gene 1.0 ST Array 10865800 - -
          Rat Genome U34 Set rc_AA817999_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21184334 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364027 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0