Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01492634_m1
          See other SLCO1B2 GE Assays ›
          Gene Symbol:
          Slco1b2
          Gene Name:
          solute carrier organic anion transporter family, member 1B2
          Gene Aliases:
          OATP-4, Oatp4, Slc21a10, Slco1b3, rlst-1
          Chromosome Location:
          Chr.4: 175814118 - 175881775 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01492634_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slco1b2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270586.1 NP_001257515.1 14 - 15 1855 83
          NM_001270587.1 NP_001257516.1 13 - 14 1861 83
          NM_031650.3 NP_113838.1 14 - 15 1960 83
          GenBank mRNA AF147740.1 - 13 - 14 1741 83
          AF208545.2 - 14 - 15 1960 83
          AF217450.2 - 13 - 14 1861 83
          AJ271682.1 - 14 - 15 1961 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001999.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slco1b2

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          xenobiotic transmembrane transport
          oligopeptide transport
          response to xenobiotic stimulus
          bile acid and bile salt transport
          response to lipopolysaccharide
          response to lipid
          response to cytokine
          xenobiotic transport
          response to peptide hormone
          response to antibiotic
          response to glucocorticoid
          organic anion transmembrane transporter activity
          bile acid transmembrane transporter activity
          oligopeptide transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P627436 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P328097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941197 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171776 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121709 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149140 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P627436 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P328097 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P328097 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P627436 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P328097 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P627436 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P328097 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17323 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70076 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45461 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92813 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12938 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10859545 - -
          Affymetrix Rat Gene 1.0 ST Array 10859529 - -
          Affymetrix Rat Gene 1.0 ST Array 10859536 - -
          Affymetrix Rat Gene 1.0 ST Array 10859542 - -
          Affymetrix Rat Gene 1.0 ST Array 10859530 - -
          Affymetrix Rat Gene 1.0 ST Array 10859533 - -
          Affymetrix Rat Gene 1.0 ST Array 10859540 - -
          Affymetrix Rat Gene 1.0 ST Array 10859537 - -
          Affymetrix Rat Gene 1.0 ST Array 10859528 - -
          Affymetrix Rat Gene 1.0 ST Array 10859539 - -
          Affymetrix Rat Gene 1.0 ST Array 10859535 - -
          Affymetrix Rat Gene 1.0 ST Array 10859531 - -
          Affymetrix Rat Gene 1.0 ST Array 10859538 - -
          Affymetrix Rat Gene 1.0 ST Array 10859532 - -
          Affymetrix Rat Gene 1.0 ST Array 10859543 - -
          Affymetrix Rat Gene 1.0 ST Array 10859534 - -
          Affymetrix Rat Gene 1.0 ST Array 10859541 - -
          Affymetrix Rat Gene 1.0 ST Array 10859546 - -
          Affymetrix Rat Gene 1.0 ST Array 10859544 - -
          Affymetrix Rat Gene 1.1 ST Array 10859527 - -
          Rat Genome 230 2.0 Array 1369746_PM_a_at - -
          Rat Genome 230 2.0 Array 1387679_at - -
          Rat Genome 230 2.0 Array 1387679_PM_at - -
          Rat Genome 230 2.0 Array 1369746_a_at - -
          Affymetrix HT Rat Focus Array 1369746_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387679_PM_at - -
          Affymetrix HT Rat Focus Array 1387679_at - -
          Affymetrix HT Rat Focus Array 1369746_a_at - -
          Affymetrix Rat Clariom S Assay PSR0400014468.rn.1 TC0400001999.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400014467.rn.1 TC0400001999.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400014469.rn.1 TC0400001999.rn.1 RTA TC
          Rat Expression Set 230 1387679_PM_at - -
          Rat Expression Set 230 1369746_PM_a_at - -
          Rat Expression Set 230 1369746_a_at - -
          Rat Expression Set 230 1387679_at - -
          Affymetrix Rat Gene 1.0 ST Array 10859527 - -
          Rat Genome U34 Set rc_AA945533_at - -
          Rat Genome U34 Set rc_AI169779_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22293596 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361516 - -
          RatRef-12 Expression BeadChip ILMN_48348 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0