Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01494340_m1
          See other AK6 GE Assays ›
          Gene Symbol:
          Ak6 Taf9
          Gene Name:
          adenylate kinase 6
          TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor
          Gene Aliases:
          AK/ATPase
          CINAP, TAFII31, TAFII32
          Chromosome Location:
          Chr.2: 30664407 - 30676464 on Build Rnor_6.0
          Chromosome Location:
          Chr.2: 30664407 - 30668414 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01494340_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ak6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012463.3 NP_001012481.1 1 - 2 80 78
          GenBank mRNA BC089989.1 - 1 - 2 46 78
          Gene Symbol: Taf9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_184048.2 NP_908937.2 1 - 2 80 78
          GenBank mRNA CB724309.1 - 1 - 2 69 78
          CB743840.1 - 1 - 2 270 78
          FQ213120.1 - 1 - 2 63 78
          FQ215462.1 - 1 - 2 74 78
          FQ229736.1 - 1 - 2 73 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000263.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the BC089989.1, NM_001012463.3, NM_184048.2 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          coenzyme A biosynthetic process
          phosphorylation
          nucleotide phosphorylation
          DNA-templated transcription, initiation
          positive regulation of cell growth
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          histone H3 acetylation
          positive regulation of transcription by RNA polymerase II
          protein stabilization
          positive regulation of response to cytokine stimulus
          response to interleukin-1
          response to L-glutamate
          adenylate kinase activity
          dephospho-CoA kinase activity
          ATP binding
          ATP hydrolysis activity
          transcription cis-regulatory region binding
          p53 binding
          DNA binding
          transcription coactivator activity
          histone acetyltransferase activity
          protein binding
          protein heterodimerization activity
          C2H2 zinc finger domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0200001486.rn.1 TC0200000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200001497.rn.1 TC0200000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200001477.rn.1 TC0200000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200001496.rn.1 TC0200000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200001476.rn.1 TC0200000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200001495.rn.1 TC0200000263.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0