Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01496225_g1
          See other MED11 GE Assays ›
          Gene Symbol:
          Med11
          Gene Name:
          mediator complex subunit 11
          Gene Aliases:
          RGD1563202
          Chromosome Location:
          Chr.10: 57057272 - 57059415 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01496225_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Med11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105799.1 NP_001099269.1 3 - 4 360 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000963.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          protein ubiquitination
          transcription coregulator activity
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P993464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462468 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P993464 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P993464 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P993464 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P993464 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10735220 - -
          Affymetrix Rat Gene 1.0 ST Array 10735218 - -
          Affymetrix Rat Gene 1.0 ST Array 10735219 - -
          Affymetrix Rat Gene 1.1 ST Array 10735217 - -
          Rat Genome 230 2.0 Array 1388621_PM_at - -
          Rat Genome 230 2.0 Array 1388621_at - -
          Affymetrix HT Rat Focus Array 1388621_PM_at - -
          Affymetrix HT Rat Focus Array 1388621_at - -
          Affymetrix Rat Clariom S Assay PSR1000008511.rn.1 TC1000000963.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008507.rn.1 TC1000000963.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008513.rn.1 TC1000000963.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008512.rn.1 TC1000000963.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000008514.rn.1 TC1000000963.rn.1 RTA TC
          Rat Expression Set 230 1388621_PM_at - -
          Rat Expression Set 230 1388621_at - -
          Affymetrix Rat Gene 1.0 ST Array 10735217 - -
          Rat Genome U34 Set rc_AI072195_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21838178 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354936 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0