Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01497022_gH
          See other STAT5B GE Assays ›
          Gene Symbol:
          Stat5b
          Gene Name:
          signal transducer and activator of transcription 5B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 88686207 - 88712313 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01497022_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stat5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022380.1 NP_071775.1 2342 60
          GenBank mRNA X91988.1 - 17 - 18 2376 60
          X97541.1 - 2342 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003812.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stat5b

          Gene Ontology Categories:

          Function(s) Process(es)

          allantoin metabolic process
          luteinization
          response to hypoxia
          natural killer cell differentiation
          liver development
          citrate metabolic process
          2-oxoglutarate metabolic process
          succinate metabolic process
          oxaloacetate metabolic process
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          isoleucine metabolic process
          valine metabolic process
          creatine metabolic process
          fatty acid metabolic process
          acute-phase response
          signal transduction
          receptor signaling pathway via JAK-STAT
          sex differentiation
          female pregnancy
          lactation
          positive regulation of cell population proliferation
          regulation of steroid metabolic process
          cytokine-mediated signaling pathway
          taurine metabolic process
          lipid storage
          regulation of cell adhesion
          regulation of epithelial cell differentiation
          response to estradiol
          response to lipopolysaccharide
          positive regulation of natural killer cell proliferation
          positive regulation of natural killer cell differentiation
          cellular response to hormone stimulus
          T cell differentiation in thymus
          prolactin signaling pathway
          regulation of multicellular organism growth
          positive regulation of multicellular organism growth
          positive regulation of activated T cell proliferation
          progesterone metabolic process
          T cell homeostasis
          negative regulation of apoptotic process
          response to peptide hormone
          response to ethanol
          positive regulation of B cell differentiation
          positive regulation of gamma-delta T cell differentiation
          positive regulation of lymphocyte differentiation
          negative regulation of erythrocyte differentiation
          positive regulation of mitotic cell cycle
          positive regulation of transcription by RNA polymerase II
          positive regulation of natural killer cell mediated cytotoxicity
          creatinine metabolic process
          development of secondary female sexual characteristics
          development of secondary male sexual characteristics
          Peyer's patch development
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          positive regulation of cellular component movement
          growth hormone receptor signaling pathway via JAK-STAT
          response to interleukin-2
          response to interleukin-4
          response to interleukin-15
          cellular response to growth factor stimulus
          cellular response to epidermal growth factor stimulus
          RNA polymerase II core promoter sequence-specific DNA binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          DNA-binding transcription factor activity
          protein binding
          protein phosphatase binding
          glucocorticoid receptor binding
          sequence-specific DNA binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30158 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93153 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117686 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12209 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12209 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12209 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12209 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700502882 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7519 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49537 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1022 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104430 - -
          Agilent Rat cDNA Microarray (G4105A) 700502882 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10747498 - -
          Affymetrix Rat Gene 1.0 ST Array 10747502 - -
          Affymetrix Rat Gene 1.0 ST Array 10747499 - -
          Affymetrix Rat Gene 1.0 ST Array 10747500 - -
          Affymetrix Rat Gene 1.0 ST Array 10747496 - -
          Affymetrix Rat Gene 1.0 ST Array 10747497 - -
          Affymetrix Rat Gene 1.0 ST Array 10747505 - -
          Affymetrix Rat Gene 1.0 ST Array 10747501 - -
          Affymetrix Rat Gene 1.0 ST Array 10747504 - -
          Affymetrix Rat Gene 1.0 ST Array 10747495 - -
          Affymetrix Rat Gene 1.1 ST Array 10747494 - -
          Rat Genome 230 2.0 Array 1374611_at - -
          Rat Genome 230 2.0 Array 1374611_PM_at - -
          Rat Genome 230 2.0 Array 1384357_at - -
          Rat Genome 230 2.0 Array 1384357_PM_at - -
          Rat Genome 230 2.0 Array 1387876_PM_at - -
          Rat Genome 230 2.0 Array 1396449_PM_at - -
          Rat Genome 230 2.0 Array 1387876_at - -
          Rat Genome 230 2.0 Array 1396449_at - -
          Affymetrix HT Rat Focus Array 1387876_PM_at - -
          Affymetrix HT Rat Focus Array 1387876_at - -
          Affymetrix Rat Clariom S Assay PSR1000034224.rn.1 TC1000003812.rn.1 RTA TC
          Rat Expression Set 230 1387876_PM_at - -
          Rat Expression Set 230 1387876_at - -
          Affymetrix Rat Gene 1.0 ST Array 10747494 - -
          Rat Genome U34 Set X91988_at - -
          Rat Genome U34 Set X91988_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22241704 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371836 - -
          RatRef-12 Expression BeadChip ILMN_53449 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0