Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01497024_m1
          See other STAT5B GE Assays ›
          Gene Symbol:
          Stat5b
          Gene Name:
          signal transducer and activator of transcription 5B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 88686207 - 88712313 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01497024_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stat5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022380.1 NP_071775.1 241 110
          GenBank mRNA X91988.1 - 1 - 2 275 110
          X97541.1 - 241 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003812.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stat5b

          Gene Ontology Categories:

          Function(s) Process(es)

          allantoin metabolic process
          luteinization
          response to hypoxia
          natural killer cell differentiation
          liver development
          citrate metabolic process
          2-oxoglutarate metabolic process
          succinate metabolic process
          oxaloacetate metabolic process
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          isoleucine metabolic process
          valine metabolic process
          creatine metabolic process
          fatty acid metabolic process
          acute-phase response
          signal transduction
          receptor signaling pathway via JAK-STAT
          sex differentiation
          female pregnancy
          lactation
          positive regulation of cell population proliferation
          regulation of steroid metabolic process
          cytokine-mediated signaling pathway
          taurine metabolic process
          lipid storage
          regulation of cell adhesion
          regulation of epithelial cell differentiation
          response to estradiol
          response to lipopolysaccharide
          positive regulation of natural killer cell proliferation
          positive regulation of natural killer cell differentiation
          cellular response to hormone stimulus
          T cell differentiation in thymus
          prolactin signaling pathway
          regulation of multicellular organism growth
          positive regulation of multicellular organism growth
          positive regulation of activated T cell proliferation
          progesterone metabolic process
          T cell homeostasis
          negative regulation of apoptotic process
          response to peptide hormone
          response to ethanol
          positive regulation of B cell differentiation
          positive regulation of gamma-delta T cell differentiation
          positive regulation of lymphocyte differentiation
          negative regulation of erythrocyte differentiation
          positive regulation of mitotic cell cycle
          positive regulation of transcription by RNA polymerase II
          positive regulation of natural killer cell mediated cytotoxicity
          creatinine metabolic process
          development of secondary female sexual characteristics
          development of secondary male sexual characteristics
          Peyer's patch development
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          positive regulation of cellular component movement
          growth hormone receptor signaling pathway via JAK-STAT
          response to interleukin-2
          response to interleukin-4
          response to interleukin-15
          cellular response to growth factor stimulus
          cellular response to epidermal growth factor stimulus
          RNA polymerase II core promoter sequence-specific DNA binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          DNA-binding transcription factor activity
          protein binding
          protein phosphatase binding
          glucocorticoid receptor binding
          sequence-specific DNA binding
          protein dimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1000034248.rn.1 TC1000003812.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034238.rn.1 TC1000003812.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034239.rn.1 TC1000003812.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034241.rn.1 TC1000003812.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034236.rn.1 TC1000003812.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034240.rn.1 TC1000003812.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034243.rn.1 TC1000003812.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034237.rn.1 TC1000003812.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0