Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01497988_g1
          See other GAS5 GE Assays ›
          Gene Symbol:
          Gas5
          Gene Name:
          growth arrest specific 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 78852523 - 78855844 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01497988_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gas5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002704.1 9 - 10 295 64
          GenBank mRNA U77829.1 - 9 - 10 295 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000605.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P552441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190808 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P552441 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P552441 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P552441 - -
          Array Name Probe IDs Targets Target Type
          Rat Genome 230 2.0 Array 1373499_at - -
          Rat Genome 230 2.0 Array 1392818_PM_at - -
          Rat Genome 230 2.0 Array 1373499_PM_at - -
          Rat Genome 230 2.0 Array 1392818_at - -
          Affymetrix HT Rat Focus Array 1373499_at - -
          Affymetrix HT Rat Focus Array 1373499_PM_at - -
          Affymetrix HT Rat Focus Array 1392818_PM_at - -
          Affymetrix HT Rat Focus Array 1392818_at - -
          Affymetrix Rat Clariom S Assay PSR1300004172.rn.1 TC1300000605.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004175.rn.1 TC1300000605.rn.1 RTA TC
          Rat Expression Set 230 1373499_PM_at - -
          Rat Expression Set 230 1373499_at - -
          Rat Expression Set 230 1392818_at - -
          Rat Expression Set 230 1392818_PM_at - -
          Rat Genome U34 Set U77829mRNA_s_at - -
          Rat Genome U34 Set U77829mRNA_i_at - -
          Rat Genome U34 Set rc_AI111599_at - -
          Rat Genome U34 Set rc_AI111599_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21131190 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0