Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01498101_m1
          See other CD163L1 GE Assays ›
          Gene Symbol:
          Cd163l1
          Gene Name:
          CD163 molecule-like 1
          Gene Aliases:
          RGD1307378
          Chromosome Location:
          Chr.1: 212695567 - 212706986 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01498101_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd163l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106312.1 NP_001099782.1 2 - 3 693 119
          XM_008759975.1 XP_008758197.1 4 - 5 717 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002842.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor-mediated endocytosis
          scavenger receptor activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032762 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557232 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85149 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032762 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112630 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112630 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032762 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032762 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112630 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P229403 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10712038 - -
          Affymetrix Rat Gene 1.0 ST Array 10712037 - -
          Affymetrix Rat Gene 1.0 ST Array 10712035 - -
          Affymetrix Rat Gene 1.0 ST Array 10712039 - -
          Affymetrix Rat Gene 1.0 ST Array 10712043 - -
          Affymetrix Rat Gene 1.0 ST Array 10712042 - -
          Affymetrix Rat Gene 1.0 ST Array 10712040 - -
          Affymetrix Rat Gene 1.0 ST Array 10712036 - -
          Affymetrix Rat Gene 1.0 ST Array 10712041 - -
          Affymetrix Rat Gene 1.0 ST Array 10712045 - -
          Affymetrix Rat Gene 1.0 ST Array 10712046 - -
          Affymetrix Rat Gene 1.1 ST Array 10712034 - -
          Affymetrix Rat Clariom S Assay PSR0100022453.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022455.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022456.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022445.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022443.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022446.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022448.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022452.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022449.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022447.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022444.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022451.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022450.rn.1 TC0100002842.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10712034 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc