Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01498525_m1
          See other ZFP428 GE Assays ›
          Gene Symbol:
          Zfp428
          Gene Name:
          zinc finger protein 428
          Gene Aliases:
          RGD1564862
          Chromosome Location:
          Chr.1: 81371324 - 81382084 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01498525_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp428
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108476.1 NP_001101946.1 2 - 3 625 107
          XM_008758954.1 XP_008757176.1 1 - 2 1368 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001044.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P431154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791173 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55276 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20190 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P431154 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P431154 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P431154 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P431154 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46518 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704970 - -
          Affymetrix Rat Gene 1.0 ST Array 10704973 - -
          Affymetrix Rat Gene 1.0 ST Array 10704971 - -
          Affymetrix Rat Gene 1.1 ST Array 10704969 - -
          Rat Genome 230 2.0 Array 1389099_PM_at - -
          Rat Genome 230 2.0 Array 1389099_at - -
          Rat Genome 230 2.0 Array 1378975_PM_at - -
          Rat Genome 230 2.0 Array 1378975_at - -
          Affymetrix HT Rat Focus Array 1389099_at - -
          Affymetrix HT Rat Focus Array 1378975_at - -
          Affymetrix HT Rat Focus Array 1378975_PM_at - -
          Affymetrix HT Rat Focus Array 1389099_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100007379.rn.1 TC0100001044.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007380.rn.1 TC0100001044.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007377.rn.1 TC0100001044.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007382.rn.1 TC0100001044.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007381.rn.1 TC0100001044.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007383.rn.1 TC0100001044.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007378.rn.1 TC0100001044.rn.1 RTA TC
          Rat Expression Set 230 1389099_PM_at - -
          Rat Expression Set 230 1389099_at - -
          Rat Expression Set 230 1378975_PM_at - -
          Rat Expression Set 230 1378975_at - -
          Affymetrix Rat Gene 1.0 ST Array 10704969 - -
          Rat Genome U34 Set rc_AI009137_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21399189 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372727 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0