Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01498920_g1
          See other ZFP36L2 GE Assays ›
          Gene Symbol:
          Zfp36l2 LOC100911319
          Gene Name:
          zinc finger protein 36, C3H type-like 2
          zinc finger protein 36, C3H1 type-like 2-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.6: 7417416 - 7423484 on Build Rnor_6.0
          Chromosome Location:
          Chr.7: 2350068 - 2354108 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01498920_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp36l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001036626.1 NP_001031703.1 1 - 2 284 71
          GenBank mRNA CV112816.1 - 1 - 2 284 71
          Gene Symbol: LOC100911319
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003750261.1 XP_003750309.1 1 - 2 284 71

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp36l2
          Click here to find siRNAs for Gene LOC100911319

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          mRNA catabolic process
          hemopoiesis
          T cell differentiation in thymus
          somatic stem cell population maintenance
          regulation of mRNA stability
          somatic stem cell division
          definitive hemopoiesis
          positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          negative regulation of stem cell differentiation
          RNA binding
          mRNA 3'-UTR AU-rich region binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P239765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456335 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30301 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P239765 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P239765 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P239765 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P115932 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P373071 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103614 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10888151 - -
          Affymetrix Rat Gene 1.0 ST Array 10888152 - -
          Affymetrix Rat Gene 1.1 ST Array 10888150 - -
          Rat Genome 230 2.0 Array 1373106_PM_at - -
          Rat Genome 230 2.0 Array 1375344_PM_at - -
          Rat Genome 230 2.0 Array 1373106_at - -
          Rat Genome 230 2.0 Array 1375344_at - -
          Rat Genome 230 2.0 Array 1389851_at - -
          Rat Genome 230 2.0 Array 1389851_PM_at - -
          Affymetrix HT Rat Focus Array 1389851_at - -
          Affymetrix HT Rat Focus Array 1373106_PM_at - -
          Affymetrix HT Rat Focus Array 1389851_PM_at - -
          Affymetrix HT Rat Focus Array 1373106_at - -
          Rat Expression Set 230 1373106_PM_at - -
          Rat Expression Set 230 1375344_PM_at - -
          Rat Expression Set 230 1373106_at - -
          Rat Expression Set 230 1375344_at - -
          Rat Expression Set 230 1389851_at - -
          Rat Expression Set 230 1389851_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10888150 - -
          Rat Genome U34 Set rc_AA943730_at - -
          Rat Genome U34 Set rc_AI178069_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21577714 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369812 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0