Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499101_g1
          See other RPS7 GE Assays ›
          Gene Symbol:
          Rps7 LOC100362830
          Gene Name:
          ribosomal protein S7
          ribosomal protein S7-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.6: 47899525 - 47904389 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 169021515 - 169022178 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01499101_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031570.2 NP_113758.2 5 - 6 395 130
          GenBank mRNA AA819435.1 - 2 - 3 130
          BC060557.1 - 5 - 6 399 130
          BC168722.1 - 4 - 5 379 130
          FQ210052.1 - 5 - 6 395 130
          FQ211139.1 - 5 - 6 395 130
          FQ212121.1 - 5 - 6 395 130
          FQ217452.1 - 5 - 6 395 130
          FQ219950.1 - 5 - 6 395 130
          FQ220886.1 - 5 - 6 443 130
          FQ221065.1 - 5 - 6 395 130
          FQ221374.1 - 5 - 6 395 130
          FQ221761.1 - 5 - 6 395 130
          FQ221932.1 - 5 - 6 398 130
          FQ222169.1 - 5 - 6 395 130
          FQ222484.1 - 5 - 6 395 130
          FQ222534.1 - 5 - 6 393 130
          FQ222918.1 - 5 - 6 395 130
          FQ222969.1 - 2 - 3 146 130
          FQ222996.1 - 5 - 6 395 130
          FQ223010.1 - 5 - 6 395 130
          FQ223103.1 - 5 - 6 395 130
          FQ223290.1 - 5 - 6 395 130
          FQ223392.1 - 5 - 6 395 130
          FQ223426.1 - 5 - 6 395 130
          FQ223491.1 - 5 - 6 395 130
          FQ228541.1 - 5 - 6 411 130
          FQ229000.1 - 5 - 6 130
          X53377.1 - 5 - 6 395 130
          Gene Symbol: LOC100362830
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002729576.4 XP_002729622.2 1 395 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003499.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps7
          Click here to find siRNAs for Gene LOC100362830

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          cytoplasmic translation
          rRNA processing
          biological_process
          cell differentiation
          ribosomal small subunit biogenesis
          molecular_function
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0600024747.rn.1 TC0600003499.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024744.rn.1 TC0600003499.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024748.rn.1 TC0600003499.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024749.rn.1 TC0600003499.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024745.rn.1 TC0600003499.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0