Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01500033_gH
          See other RPLP0 GE Assays ›
          Gene Symbol:
          Rplp0
          Gene Name:
          ribosomal protein, large, P0
          Gene Aliases:
          Arbp
          Chromosome Location:
          Chr.12: 46791528 - 46794797 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01500033_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rplp0
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022402.2 NP_071797.1 5 - 6 728 92
          GenBank mRNA BC062028.1 - 4 - 5 697 92
          BP465362.1 - 2 - 3 92
          DN931898.1 - 4 - 5 746 92
          FQ209603.1 - 5 - 6 728 92
          FQ209620.1 - 4 - 5 713 92
          FQ209649.1 - 5 - 6 92
          FQ209757.1 - 5 - 6 729 92
          FQ210253.1 - 5 - 6 728 92
          FQ210412.1 - 5 - 6 728 92
          FQ211152.1 - 5 - 6 728 92
          FQ211263.1 - 5 - 6 727 92
          FQ212554.1 - 5 - 6 728 92
          FQ212743.1 - 5 - 6 728 92
          FQ212875.1 - 5 - 6 728 92
          FQ212927.1 - 5 - 6 729 92
          FQ213085.1 - 5 - 6 728 92
          FQ213427.1 - 5 - 6 728 92
          FQ213510.1 - 5 - 6 728 92
          FQ213712.1 - 5 - 6 728 92
          FQ214087.1 - 5 - 6 728 92
          FQ214354.1 - 5 - 6 728 92
          FQ214553.1 - 5 - 6 729 92
          FQ214850.1 - 5 - 6 728 92
          FQ214997.1 - 5 - 6 728 92
          FQ215548.1 - 5 - 6 92
          FQ215662.1 - 5 - 6 728 92
          FQ215682.1 - 5 - 6 728 92
          FQ215870.1 - 5 - 6 728 92
          FQ216120.1 - 5 - 6 729 92
          FQ216296.1 - 5 - 6 92
          FQ216389.1 - 5 - 6 729 92
          FQ216495.1 - 5 - 6 728 92
          FQ216618.1 - 5 - 6 728 92
          FQ216655.1 - 5 - 6 729 92
          FQ216733.1 - 5 - 6 730 92
          FQ216774.1 - 5 - 6 728 92
          FQ216813.1 - 5 - 6 92
          FQ216851.1 - 5 - 6 728 92
          FQ216913.1 - 5 - 6 729 92
          FQ217406.1 - 5 - 6 728 92
          FQ218561.1 - 5 - 6 728 92
          FQ218943.1 - 5 - 6 728 92
          FQ219025.1 - 5 - 6 92
          FQ219184.1 - 5 - 6 732 92
          FQ219389.1 - 5 - 6 728 92
          FQ219457.1 - 5 - 6 728 92
          FQ219678.1 - 5 - 6 728 92
          FQ219815.1 - 5 - 6 728 92
          FQ219817.1 - 5 - 6 728 92
          FQ219837.1 - 5 - 6 728 92
          FQ219855.1 - 5 - 6 728 92
          FQ219923.1 - 5 - 6 728 92
          FQ220008.1 - 5 - 6 728 92
          FQ220051.1 - 5 - 6 728 92
          FQ220077.1 - 5 - 6 728 92
          FQ220115.1 - 5 - 6 728 92
          FQ220181.1 - 5 - 6 728 92
          FQ220201.1 - 5 - 6 726 92
          FQ220213.1 - 5 - 6 728 92
          FQ220247.1 - 5 - 6 729 92
          FQ220282.1 - 5 - 6 730 92
          FQ220284.1 - 5 - 6 728 92
          FQ220338.1 - 5 - 6 728 92
          FQ220345.1 - 5 - 6 729 92
          FQ220370.1 - 5 - 6 728 92
          FQ220420.1 - 5 - 6 729 92
          FQ220433.1 - 5 - 6 725 92
          FQ220444.1 - 5 - 6 728 92
          FQ220445.1 - 5 - 6 724 92
          FQ220449.1 - 5 - 6 728 92
          FQ220500.1 - 5 - 6 728 92
          FQ220514.1 - 5 - 6 728 92
          FQ220527.1 - 5 - 6 728 92
          FQ220533.1 - 5 - 6 760 92
          FQ220542.1 - 5 - 6 728 92
          FQ220545.1 - 5 - 6 728 92
          FQ220634.1 - 5 - 6 728 92
          FQ220654.1 - 5 - 6 728 92
          FQ220663.1 - 5 - 6 728 92
          FQ220684.1 - 5 - 6 729 92
          FQ220690.1 - 5 - 6 730 92
          FQ220769.1 - 5 - 6 728 92
          FQ220787.1 - 5 - 6 728 92
          FQ220801.1 - 5 - 6 728 92
          FQ220833.1 - 5 - 6 728 92
          FQ220838.1 - 5 - 6 728 92
          FQ220888.1 - 5 - 6 728 92
          FQ220932.1 - 5 - 6 729 92
          FQ221146.1 - 5 - 6 728 92
          FQ221380.1 - 5 - 6 728 92
          FQ221533.1 - 5 - 6 729 92
          FQ221556.1 - 5 - 6 726 92
          FQ222171.1 - 5 - 6 728 92
          FQ222181.1 - 5 - 6 728 92
          FQ222327.1 - 5 - 6 728 92
          FQ223487.1 - 5 - 6 728 92
          FQ224547.1 - 5 - 6 728 92
          FQ224834.1 - 5 - 6 728 92
          FQ224846.1 - 5 - 6 728 92
          FQ225049.1 - 5 - 6 728 92
          FQ225170.1 - 5 - 6 728 92
          FQ225608.1 - 5 - 6 728 92
          FQ225654.1 - 5 - 6 730 92
          FQ225886.1 - 5 - 6 728 92
          FQ226306.1 - 5 - 6 92
          FQ227381.1 - 5 - 6 729 93
          FQ227522.1 - 5 - 6 723 92
          FQ227719.1 - 5 - 6 728 92
          FQ227837.1 - 5 - 6 728 92
          FQ227937.1 - 5 - 6 729 92
          FQ227941.1 - 5 - 6 728 92
          FQ227942.1 - 5 - 6 848 92
          FQ228995.1 - 5 - 6 728 92
          FQ228996.1 - 5 - 6 728 92
          FQ229139.1 - 5 - 6 730 92
          FQ229164.1 - 5 - 6 729 92
          FQ229241.1 - 5 - 6 728 92
          FQ229244.1 - 5 - 6 728 92
          FQ229272.1 - 5 - 6 728 92
          FQ229273.1 - 5 - 6 729 92
          FQ229303.1 - 5 - 6 728 92
          FQ229337.1 - 5 - 6 728 92
          FQ229352.1 - 5 - 6 728 92
          FQ229365.1 - 5 - 6 728 92
          FQ229397.1 - 5 - 6 728 92
          FQ229408.1 - 5 - 6 729 92
          FQ229433.1 - 5 - 6 728 92
          FQ229448.1 - 5 - 6 728 92
          FQ229521.1 - 5 - 6 728 92
          FQ229547.1 - 5 - 6 728 92
          FQ229554.1 - 5 - 6 92
          FQ229566.1 - 5 - 6 724 92
          FQ229610.1 - 5 - 6 728 92
          FQ229626.1 - 5 - 6 725 92
          FQ229726.1 - 5 - 6 728 92
          FQ229740.1 - 5 - 6 728 92
          FQ229742.1 - 5 - 6 728 92
          FQ229808.1 - 5 - 6 728 92
          FQ229815.1 - 5 - 6 728 92
          FQ229822.1 - 5 - 6 728 92
          FQ229830.1 - 5 - 6 730 92
          FQ229883.1 - 5 - 6 730 92
          FQ229929.1 - 5 - 6 728 92
          FQ229932.1 - 3 - 4 626 92
          FQ229952.1 - 5 - 6 728 92
          FQ229974.1 - 5 - 6 728 92
          FQ230005.1 - 5 - 6 728 92
          FQ230055.1 - 5 - 6 727 92
          FQ230127.1 - 5 - 6 728 92
          FQ230267.1 - 5 - 6 731 92
          FQ230943.1 - 5 - 6 728 92
          FQ231445.1 - 5 - 6 730 92
          FQ231525.1 - 5 - 6 728 92
          FQ231591.1 - 5 - 6 730 92
          FQ231733.1 - 5 - 6 729 92
          FQ231897.1 - 5 - 6 730 92
          FQ231967.1 - 5 - 6 728 92
          FQ232029.1 - 5 - 6 729 92
          FQ232154.1 - 5 - 6 728 92
          FQ232165.1 - 5 - 6 728 92
          FQ232310.1 - 5 - 6 727 92
          FQ232319.1 - 5 - 6 729 92
          FQ232697.1 - 5 - 6 730 92
          FQ232767.1 - 5 - 6 728 92
          FQ232951.1 - 5 - 6 728 92
          FQ233152.1 - 5 - 6 727 92
          FQ233377.1 - 5 - 6 728 92
          FQ233473.1 - 5 - 6 728 92
          FQ233732.1 - 5 - 6 728 92
          FQ233737.1 - 5 - 6 728 92
          FQ233808.1 - 5 - 6 728 92
          FQ234195.1 - 5 - 6 728 92
          FQ234341.1 - 5 - 6 728 92
          FQ234613.1 - 5 - 6 728 92
          FQ234733.1 - 5 - 6 728 92
          FQ234735.1 - 5 - 6 618 92
          FQ235063.1 - 5 - 6 728 92
          FQ235079.1 - 5 - 6 728 92
          FQ235122.1 - 5 - 6 728 92
          FQ235259.1 - 5 - 6 728 92
          X15096.1 - 5 - 6 728 92
          Z29530.1 - 4 - 5 684 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001737.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection GE Endogenous Controls Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rplp0

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          ribosome biogenesis
          cellular response to interleukin-4
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047118 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797023 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047118 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14914 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047118 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14914 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14914 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047118 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P544108 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701592887 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700530845 - -
          Agilent Rat cDNA Microarray (G4105A) 700530845 - -
          Agilent Rat cDNA Microarray (G4105A) 701592887 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10762659 - -
          Affymetrix Rat Gene 1.0 ST Array 10762664 - -
          Affymetrix Rat Gene 1.0 ST Array 10762663 - -
          Affymetrix Rat Gene 1.0 ST Array 10731733 - -
          Affymetrix Rat Gene 1.0 ST Array 10762661 - -
          Affymetrix Rat Gene 1.0 ST Array 10762662 - -
          Affymetrix Rat Gene 1.0 ST Array 10717134 - -
          Affymetrix Rat Gene 1.0 ST Array 10762660 - -
          Affymetrix Rat Gene 1.1 ST Array 10762658 - -
          Affymetrix Rat Gene 1.1 ST Array 10717132 - -
          Affymetrix Rat Gene 1.1 ST Array 10731732 - -
          Rat Genome 230 2.0 Array 1367560_at - -
          Rat Genome 230 2.0 Array 1367560_PM_at - -
          Affymetrix HT Rat Focus Array 1367560_PM_at - -
          Affymetrix HT Rat Focus Array 1367560_at - -
          Affymetrix Rat Clariom S Assay PSR1200014845.rn.1 TC1200001737.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200014847.rn.1 TC1200001737.rn.1 RTA TC
          Rat Expression Set 230 1367560_at - -
          Rat Expression Set 230 1367560_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10762658 - -
          Affymetrix Rat Gene 1.0 ST Array 10731732 - -
          Rat Genome U34 Set X15096cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20882705 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357284 - -
          RatRef-12 Expression BeadChip ILMN_67119 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline