Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01501048_m1
          See other ASB4 GE Assays ›
          Gene Symbol:
          Asb4
          Gene Name:
          ankyrin repeat and SOCS box-containing 4
          Gene Aliases:
          Asb-4
          Chromosome Location:
          Chr.4: 30461686 - 30501340 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01501048_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Asb4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024318.1 NP_001019489.1 3 - 4 987 77
          XM_008762714.1 XP_008760936.1 4 - 5 1393 77
          GenBank mRNA AY577765.1 - 3 - 4 987 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000318.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Asb4

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular signal transduction
          protein autoubiquitination
          positive regulation of vasculogenesis
          ubiquitin-protein transferase activity
          enzyme binding
          ubiquitin protein ligase binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P590244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316063 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39394 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P590244 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P590244 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P590244 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P590244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72115 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76070 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76802 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24088 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853638 - -
          Affymetrix Rat Gene 1.0 ST Array 10853639 - -
          Affymetrix Rat Gene 1.0 ST Array 10853642 - -
          Affymetrix Rat Gene 1.0 ST Array 10853643 - -
          Affymetrix Rat Gene 1.0 ST Array 10853640 - -
          Affymetrix Rat Gene 1.1 ST Array 10853637 - -
          Affymetrix Rat Clariom S Assay PSR0400002383.rn.1 TC0400000318.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400002384.rn.1 TC0400000318.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10853637 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22234958 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0