Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01501534_m1
          See other ZFP91 GE Assays ›
          Gene Symbol:
          Zfp91
          Gene Name:
          zinc finger protein 91
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 229602499 - 229640917 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01501534_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp91
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001169120.1 NP_001162591.1 9 - 10 1132 68
          XM_006231127.2 XP_006231189.1 9 - 10 1386 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007320.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of NF-kappaB-inducing kinase activity
          protein K63-linked ubiquitination
          nucleic acid binding
          ubiquitin-protein transferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P337606 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P337606 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P337606 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P337606 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P755052 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10703 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P337606 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P972929 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700480387 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700373086 - -
          Agilent Rat cDNA Microarray (G4105A) 700373086 - -
          Agilent Rat cDNA Microarray (G4105A) 700480387 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10729032 - -
          Affymetrix Rat Gene 1.0 ST Array 10729031 - -
          Affymetrix Rat Gene 1.1 ST Array 10729027 - -
          Rat Genome 230 2.0 Array 1381934_PM_at - -
          Rat Genome 230 2.0 Array 1386512_PM_at - -
          Rat Genome 230 2.0 Array 1383053_PM_x_at - -
          Rat Genome 230 2.0 Array 1383053_x_at - -
          Rat Genome 230 2.0 Array 1383052_a_at - -
          Rat Genome 230 2.0 Array 1383054_PM_at - -
          Rat Genome 230 2.0 Array 1399044_PM_at - -
          Rat Genome 230 2.0 Array 1386512_at - -
          Rat Genome 230 2.0 Array 1383463_PM_at - -
          Rat Genome 230 2.0 Array 1383052_PM_a_at - -
          Rat Genome 230 2.0 Array 1383054_at - -
          Rat Genome 230 2.0 Array 1399044_at - -
          Rat Genome 230 2.0 Array 1377105_at - -
          Rat Genome 230 2.0 Array 1383463_at - -
          Rat Genome 230 2.0 Array 1381934_at - -
          Rat Genome 230 2.0 Array 1377105_PM_at - -
          Affymetrix HT Rat Focus Array 1383053_PM_x_at - -
          Affymetrix HT Rat Focus Array 1383052_a_at - -
          Affymetrix HT Rat Focus Array 1386512_at - -
          Affymetrix HT Rat Focus Array 1399044_at - -
          Affymetrix HT Rat Focus Array 1399044_PM_at - -
          Affymetrix HT Rat Focus Array 1383054_at - -
          Affymetrix HT Rat Focus Array 1381934_at - -
          Affymetrix HT Rat Focus Array 1383054_PM_at - -
          Affymetrix HT Rat Focus Array 1381934_PM_at - -
          Affymetrix HT Rat Focus Array 1383052_PM_a_at - -
          Affymetrix HT Rat Focus Array 1386512_PM_at - -
          Affymetrix HT Rat Focus Array 1383463_PM_at - -
          Affymetrix HT Rat Focus Array 1377105_PM_at - -
          Affymetrix HT Rat Focus Array 1377105_at - -
          Affymetrix HT Rat Focus Array 1383463_at - -
          Affymetrix HT Rat Focus Array 1383053_x_at - -
          Affymetrix Rat Clariom S Assay PSR0100059234.rn.1 TC0100007320.rn.1 RTA TC
          Rat Expression Set 230 1383052_PM_a_at - -
          Rat Expression Set 230 1383053_x_at - -
          Rat Expression Set 230 1383053_PM_x_at - -
          Rat Expression Set 230 1386512_PM_at - -
          Rat Expression Set 230 1399044_at - -
          Rat Expression Set 230 1377105_at - -
          Rat Expression Set 230 1399044_PM_at - -
          Rat Expression Set 230 1386512_at - -
          Rat Expression Set 230 1383054_PM_at - -
          Rat Expression Set 230 1383054_at - -
          Rat Expression Set 230 1383052_a_at - -
          Rat Expression Set 230 1377105_PM_at - -
          Rat Expression Set 230 1381934_PM_at - -
          Rat Expression Set 230 1381934_at - -
          Rat Expression Set 230 1383463_PM_at - -
          Rat Expression Set 230 1383463_at - -
          Rat Genome U34 Set rc_AA946530_at - -
          Rat Genome U34 Set rc_AI012641_at - -
          Rat Genome U34 Set rc_AI009799_at - -
          Rat Genome U34 Set rc_AI171172_i_at - -
          Rat Genome U34 Set rc_AI171172_s_at - -
          Rat Genome U34 Set rc_AI171172_r_at - -
          Rat Genome U34 Set rc_AI104658_at - -
          Rat Genome U34 Set rc_AI176930_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0