Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01502041_m1
          See other KDM2A GE Assays ›
          Gene Symbol:
          Kdm2a
          Gene Name:
          lysine (K)-specific demethylase 2A
          Gene Aliases:
          Fbxl11
          Chromosome Location:
          Chr.1: 219566130 - 219642273 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01502041_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kdm2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108515.1 NP_001101985.1 9 - 10 1231 65
          XM_006230843.2 XP_006230905.1 9 - 10 1003 65
          XM_006230844.2 XP_006230906.1 9 - 10 1602 65
          XM_008760152.1 XP_008758374.1 8 - 9 1251 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007071.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via nonhomologous end joining
          histone H3-K36 demethylation
          zinc ion binding
          unmethylated CpG binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P440752 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23422 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7279 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7601 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77950 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67591 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129836 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036268 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P440752 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036268 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P440752 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P440752 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036268 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P929706 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P822396 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P440752 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44027 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727575 - -
          Affymetrix Rat Gene 1.0 ST Array 10727566 - -
          Affymetrix Rat Gene 1.0 ST Array 10727561 - -
          Affymetrix Rat Gene 1.0 ST Array 10727559 - -
          Affymetrix Rat Gene 1.0 ST Array 10727568 - -
          Affymetrix Rat Gene 1.0 ST Array 10727581 - -
          Affymetrix Rat Gene 1.0 ST Array 10727569 - -
          Affymetrix Rat Gene 1.0 ST Array 10727564 - -
          Affymetrix Rat Gene 1.0 ST Array 10727565 - -
          Affymetrix Rat Gene 1.0 ST Array 10727574 - -
          Affymetrix Rat Gene 1.0 ST Array 10727580 - -
          Affymetrix Rat Gene 1.0 ST Array 10727577 - -
          Affymetrix Rat Gene 1.0 ST Array 10727573 - -
          Affymetrix Rat Gene 1.0 ST Array 10727578 - -
          Affymetrix Rat Gene 1.0 ST Array 10727579 - -
          Affymetrix Rat Gene 1.0 ST Array 10727562 - -
          Affymetrix Rat Gene 1.0 ST Array 10727571 - -
          Affymetrix Rat Gene 1.0 ST Array 10727572 - -
          Affymetrix Rat Gene 1.0 ST Array 10727563 - -
          Affymetrix Rat Gene 1.0 ST Array 10727567 - -
          Affymetrix Rat Gene 1.0 ST Array 10727576 - -
          Affymetrix Rat Gene 1.0 ST Array 10727570 - -
          Affymetrix Rat Gene 1.0 ST Array 10727560 - -
          Affymetrix Rat Gene 1.1 ST Array 10727558 - -
          Rat Genome 230 2.0 Array 1399154_at - -
          Rat Genome 230 2.0 Array 1390522_PM_at - -
          Rat Genome 230 2.0 Array 1384489_PM_at - -
          Rat Genome 230 2.0 Array 1390522_at - -
          Rat Genome 230 2.0 Array 1384489_at - -
          Rat Genome 230 2.0 Array 1399154_PM_at - -
          Affymetrix HT Rat Focus Array 1384489_at - -
          Affymetrix HT Rat Focus Array 1390522_PM_at - -
          Affymetrix HT Rat Focus Array 1390522_at - -
          Affymetrix HT Rat Focus Array 1384489_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100056298.rn.1 TC0100007071.rn.1 RTA TC
          Rat Expression Set 230 1399154_PM_at - -
          Rat Expression Set 230 1399154_at - -
          Rat Expression Set 230 1390522_at - -
          Rat Expression Set 230 1390522_PM_at - -
          Rat Expression Set 230 1384489_at - -
          Rat Expression Set 230 1384489_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10727558 - -
          Rat Genome U34 Set rc_AA963170_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20788527 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355197 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0